Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127442709-127443246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0408
Entrez Gene ID
9729
Gene Name
KIAA0408
Gene Aliases
-
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127459390 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

Gene Symbol
SOGA3
Entrez Gene ID
387104
Gene Name
SOGA family member 3
Gene Aliases
C6orf174, dJ403A15.3
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127519355 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

SNP Detail

SNP ID Chromosome Location
rs974761799 Chr.6: 127443138-127443138 on Build GRCh38
rs930398597 Chr.6: 127442862-127442862 on Build GRCh38
rs896220594 Chr.6: 127443154-127443154 on Build GRCh38
rs1013726233 Chr.6: 127443117-127443117 on Build GRCh38
rs1002217811 Chr.6: 127443036-127443036 on Build GRCh38
rs531831029 Chr.6: 127443156-127443156 on Build GRCh38
rs896573259 Chr.6: 127443101-127443101 on Build GRCh38
rs909449246 Chr.6: 127442773-127442773 on Build GRCh38
rs868240848 Chr.6: 127442849-127442849 on Build GRCh38
rs560550738 Chr.6: 127442863-127442863 on Build GRCh38
rs971747231 Chr.6: 127442757-127442757 on Build GRCh38
rs1036929016 Chr.6: 127442783-127442783 on Build GRCh38
rs2326593 Chr.6: 127442940-127442940 on Build GRCh38
rs920280612 Chr.6: 127442807-127442807 on Build GRCh38
rs369411644 Chr.6: 127442996-127442996 on Build GRCh38
rs919394084 Chr.6: 127443119-127443120 on Build GRCh38
rs773663664 Chr.6: 127442939-127442940 on Build GRCh38
rs35416490 Chr.6: 127442995-127442995 on Build GRCh38
rs910268070 Chr.6: 127443204-127443204 on Build GRCh38
rs942251670 Chr.6: 127443085-127443085 on Build GRCh38
rs994281164 Chr.6: 127442798-127442798 on Build GRCh38
rs903881763 Chr.6: 127443026-127443026 on Build GRCh38
rs892376545 Chr.6: 127442947-127442947 on Build GRCh38
rs1015802317 Chr.6: 127443121-127443121 on Build GRCh38
rs986091214 Chr.6: 127443046-127443046 on Build GRCh38
rs746456710 Chr.6: 127442957-127442957 on Build GRCh38
rs572309655 Chr.6: 127442875-127442875 on Build GRCh38
rs897999964 Chr.6: 127442788-127442788 on Build GRCh38
rs896114806 Chr.6: 127442829-127442829 on Build GRCh38
rs910322506 Chr.6: 127442923-127442923 on Build GRCh38
rs942325286 Chr.6: 127442777-127442777 on Build GRCh38
rs1031219059 Chr.6: 127442987-127442987 on Build GRCh38
rs944503828 Chr.6: 127442937-127442937 on Build GRCh38
rs577194177 Chr.6: 127443042-127443042 on Build GRCh38
rs909250017 Chr.6: 127443047-127443047 on Build GRCh38
rs17205908 Chr.6: 127443160-127443160 on Build GRCh38
rs1056232423 Chr.6: 127442824-127442824 on Build GRCh38
rs189719781 Chr.6: 127443195-127443195 on Build GRCh38
rs562648755 Chr.6: 127443164-127443164 on Build GRCh38
rs1000820181 Chr.6: 127442949-127442949 on Build GRCh38
rs1036617708 Chr.6: 127443088-127443088 on Build GRCh38
rs961633156 Chr.6: 127443137-127443137 on Build GRCh38
rs2875982 Chr.6: 127442996-127442996 on Build GRCh38
rs1050414044 Chr.6: 127442771-127442771 on Build GRCh38
rs1057491467 Chr.6: 127443145-127443145 on Build GRCh38
rs945324665 Chr.6: 127442974-127442974 on Build GRCh38
rs1050108120 Chr.6: 127442907-127442907 on Build GRCh38
rs951780041 Chr.6: 127443166-127443166 on Build GRCh38
rs559810242 Chr.6: 127443196-127443196 on Build GRCh38

View Full Product Details