Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151239919-151240421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP12
Entrez Gene ID
9590
Gene Name
A-kinase anchoring protein 12
Gene Aliases
AKAP250, SSeCKS
UniGene
Hs.371240
Gene Chromosome Location
Chr.6: 151239815-151358559 on Build GRCh38
Transcripts
NM_005100.3 XM_017011517.1

SNP Detail

SNP ID Chromosome Location
rs11314996 Chr.6: 151240369-151240369 on Build GRCh38
rs549330325 Chr.6: 151240101-151240101 on Build GRCh38
rs925375332 Chr.6: 151240081-151240081 on Build GRCh38
rs373354947 Chr.6: 151240225-151240225 on Build GRCh38
rs910133873 Chr.6: 151240294-151240294 on Build GRCh38
rs998246962 Chr.6: 151240356-151240356 on Build GRCh38
rs937012925 Chr.6: 151240325-151240325 on Build GRCh38
rs907008210 Chr.6: 151240200-151240200 on Build GRCh38
rs901706755 Chr.6: 151240355-151240355 on Build GRCh38
rs553536007 Chr.6: 151240121-151240121 on Build GRCh38
rs755549158 Chr.6: 151240357-151240357 on Build GRCh38
rs370979837 Chr.6: 151240218-151240218 on Build GRCh38
rs936654393 Chr.6: 151240100-151240100 on Build GRCh38
rs765452958 Chr.6: 151240197-151240197 on Build GRCh38
rs912768318 Chr.6: 151240032-151240032 on Build GRCh38
rs527468444 Chr.6: 151239992-151239992 on Build GRCh38
rs529648882 Chr.6: 151240064-151240064 on Build GRCh38
rs893035557 Chr.6: 151240332-151240332 on Build GRCh38
rs1033366283 Chr.6: 151240376-151240376 on Build GRCh38
rs905564405 Chr.6: 151240141-151240141 on Build GRCh38
rs187669073 Chr.6: 151239988-151239988 on Build GRCh38
rs1032911243 Chr.6: 151240182-151240182 on Build GRCh38
rs1001343510 Chr.6: 151240205-151240205 on Build GRCh38
rs995164827 Chr.6: 151240235-151240235 on Build GRCh38
rs573541357 Chr.6: 151240172-151240172 on Build GRCh38
rs985670075 Chr.6: 151240286-151240286 on Build GRCh38
rs758727009 Chr.6: 151240264-151240264 on Build GRCh38
rs778278331 Chr.6: 151240265-151240265 on Build GRCh38
rs3806984 Chr.6: 151240279-151240279 on Build GRCh38
rs949868432 Chr.6: 151240119-151240119 on Build GRCh38
rs1045352641 Chr.6: 151240130-151240130 on Build GRCh38
rs373126954 Chr.6: 151240019-151240019 on Build GRCh38
rs1029754427 Chr.6: 151240371-151240371 on Build GRCh38
rs571787609 Chr.6: 151240307-151240307 on Build GRCh38
rs75070320 Chr.6: 151240367-151240369 on Build GRCh38
rs986963726 Chr.6: 151240028-151240028 on Build GRCh38
rs1055441014 Chr.6: 151240224-151240224 on Build GRCh38
rs1010194358 Chr.6: 151240332-151240332 on Build GRCh38
rs115852698 Chr.6: 151240033-151240033 on Build GRCh38
rs534402016 Chr.6: 151240324-151240324 on Build GRCh38
rs376492383 Chr.6: 151240083-151240083 on Build GRCh38
rs951741694 Chr.6: 151240025-151240025 on Build GRCh38
rs1052891850 Chr.6: 151240125-151240125 on Build GRCh38
rs975454351 Chr.6: 151240046-151240046 on Build GRCh38
rs1054269049 Chr.6: 151240328-151240329 on Build GRCh38
rs547419373 Chr.6: 151240005-151240005 on Build GRCh38
rs1003651181 Chr.6: 151240322-151240322 on Build GRCh38
rs896887524 Chr.6: 151240087-151240087 on Build GRCh38
rs913148092 Chr.6: 151240160-151240160 on Build GRCh38
rs373773468 Chr.6: 151240301-151240301 on Build GRCh38
rs554173129 Chr.6: 151240367-151240367 on Build GRCh38
rs1025771461 Chr.6: 151240254-151240254 on Build GRCh38
rs1041924660 Chr.6: 151240337-151240337 on Build GRCh38
rs995594222 Chr.6: 151239965-151239965 on Build GRCh38
rs949789355 Chr.6: 151240302-151240302 on Build GRCh38
rs116453067 Chr.6: 151239963-151239963 on Build GRCh38
rs533669143 Chr.6: 151239998-151239998 on Build GRCh38
rs574344220 Chr.6: 151240384-151240384 on Build GRCh38

View Full Product Details