Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 151349081-151349588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP12
Entrez Gene ID
9590
Gene Name
A-kinase anchoring protein 12
Gene Aliases
AKAP250, SSeCKS
UniGene
Hs.371240
Gene Chromosome Location
Chr.6: 151239815-151358559 on Build GRCh38
Transcripts
NM_005100.3 NM_144497.2 XM_005267235.2 XM_017011517.1 XM_017011518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6346899 c.1137G>T p.L379L Chr.6: 151349528-151349528 on Build GRCh38
1660468 c.800A>C p.E267A Chr.6: 151349191-151349191 on Build GRCh38
4419337 c.1021G>A p.G341R Chr.6: 151349412-151349412 on Build GRCh38
4625690 c.955G>A p.D319N Chr.6: 151349346-151349346 on Build GRCh38
4877035 c.1042G>A p.E348K Chr.6: 151349433-151349433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552053449 Chr.6: 151349109-151349109 on Build GRCh38
rs894453490 Chr.6: 151349151-151349151 on Build GRCh38
rs968912380 Chr.6: 151349559-151349559 on Build GRCh38
rs780349640 Chr.6: 151349132-151349132 on Build GRCh38
rs774580485 Chr.6: 151349557-151349557 on Build GRCh38
rs139326971 Chr.6: 151349195-151349197 on Build GRCh38
rs770815831 Chr.6: 151349432-151349432 on Build GRCh38
rs769127522 Chr.6: 151349159-151349159 on Build GRCh38
rs543491382 Chr.6: 151349468-151349468 on Build GRCh38
rs764431237 Chr.6: 151349339-151349339 on Build GRCh38
rs140465822 Chr.6: 151349218-151349218 on Build GRCh38
rs201509723 Chr.6: 151349153-151349153 on Build GRCh38
rs747963991 Chr.6: 151349283-151349283 on Build GRCh38
rs887652361 Chr.6: 151349154-151349154 on Build GRCh38
rs111963593 Chr.6: 151349196-151349198 on Build GRCh38
rs765859539 Chr.6: 151349136-151349136 on Build GRCh38
rs35341741 Chr.6: 151349555-151349555 on Build GRCh38
rs138347602 Chr.6: 151349309-151349309 on Build GRCh38
rs146232694 Chr.6: 151349450-151349450 on Build GRCh38
rs1030495101 Chr.6: 151349492-151349492 on Build GRCh38
rs925840461 Chr.6: 151349382-151349382 on Build GRCh38
rs777452665 Chr.6: 151349496-151349496 on Build GRCh38
rs771800631 Chr.6: 151349295-151349295 on Build GRCh38
rs762194634 Chr.6: 151349462-151349462 on Build GRCh38
rs778357086 Chr.6: 151349239-151349239 on Build GRCh38
rs748958004 Chr.6: 151349166-151349166 on Build GRCh38
rs914772976 Chr.6: 151349252-151349252 on Build GRCh38
rs772755313 Chr.6: 151349150-151349150 on Build GRCh38
rs73780648 Chr.6: 151349152-151349152 on Build GRCh38
rs754492565 Chr.6: 151349470-151349470 on Build GRCh38
rs201823889 Chr.6: 151349387-151349387 on Build GRCh38
rs756112250 Chr.6: 151349128-151349128 on Build GRCh38
rs935933534 Chr.6: 151349388-151349388 on Build GRCh38
rs752909918 Chr.6: 151349333-151349333 on Build GRCh38
rs770710382 Chr.6: 151349515-151349515 on Build GRCh38
rs753950821 Chr.6: 151349211-151349211 on Build GRCh38
rs888677836 Chr.6: 151349493-151349493 on Build GRCh38
rs776276200 Chr.6: 151349192-151349192 on Build GRCh38
rs200318359 Chr.6: 151349433-151349433 on Build GRCh38
rs767416781 Chr.6: 151349114-151349114 on Build GRCh38
rs769926929 Chr.6: 151349445-151349445 on Build GRCh38
rs879130934 Chr.6: 151349535-151349535 on Build GRCh38
rs746800242 Chr.6: 151349420-151349420 on Build GRCh38
rs757750832 Chr.6: 151349353-151349353 on Build GRCh38
rs781279265 Chr.6: 151349518-151349518 on Build GRCh38
rs767469845 Chr.6: 151349185-151349185 on Build GRCh38
rs777489284 Chr.6: 151349418-151349418 on Build GRCh38
rs897651119 Chr.6: 151349485-151349485 on Build GRCh38
rs748824454 Chr.6: 151349365-151349365 on Build GRCh38
rs145594196 Chr.6: 151349106-151349106 on Build GRCh38
rs778554464 Chr.6: 151349408-151349408 on Build GRCh38
rs61757550 Chr.6: 151349203-151349203 on Build GRCh38
rs773208658 Chr.6: 151349304-151349304 on Build GRCh38
rs768378901 Chr.6: 151349402-151349404 on Build GRCh38
rs369207583 Chr.6: 151349527-151349527 on Build GRCh38
rs376048143 Chr.6: 151349463-151349463 on Build GRCh38
rs758837249 Chr.6: 151349338-151349338 on Build GRCh38
rs753153086 Chr.6: 151349219-151349219 on Build GRCh38
rs747736451 Chr.6: 151349411-151349411 on Build GRCh38
rs751348949 Chr.6: 151349108-151349108 on Build GRCh38
rs147375031 Chr.6: 151349554-151349554 on Build GRCh38
rs145689161 Chr.6: 151349243-151349243 on Build GRCh38
rs147724239 Chr.6: 151349124-151349124 on Build GRCh38
rs200570348 Chr.6: 151349459-151349459 on Build GRCh38
rs561110627 Chr.6: 151349412-151349412 on Build GRCh38
rs779489135 Chr.6: 151349140-151349140 on Build GRCh38
rs774177804 Chr.6: 151349174-151349174 on Build GRCh38
rs746811837 Chr.6: 151349501-151349501 on Build GRCh38
rs778464669 Chr.6: 151349480-151349480 on Build GRCh38
rs755310817 Chr.6: 151349217-151349217 on Build GRCh38
rs764470310 Chr.6: 151349253-151349253 on Build GRCh38
rs748510944 Chr.6: 151349143-151349143 on Build GRCh38
rs754707791 Chr.6: 151349340-151349340 on Build GRCh38
rs751901844 Chr.6: 151349306-151349306 on Build GRCh38
rs369573088 Chr.6: 151349358-151349358 on Build GRCh38
rs139227581 Chr.6: 151349451-151349451 on Build GRCh38
rs768281837 Chr.6: 151349168-151349168 on Build GRCh38
rs367745681 Chr.6: 151349221-151349221 on Build GRCh38
rs149525947 Chr.6: 151349490-151349490 on Build GRCh38
rs764352757 Chr.6: 151349458-151349458 on Build GRCh38
rs753951618 Chr.6: 151349138-151349138 on Build GRCh38
rs779463041 Chr.6: 151349444-151349444 on Build GRCh38
rs199669098 Chr.6: 151349307-151349307 on Build GRCh38
rs372605345 Chr.6: 151349261-151349261 on Build GRCh38
rs747406627 Chr.6: 151349242-151349242 on Build GRCh38
rs149211517 Chr.6: 151349139-151349139 on Build GRCh38
rs759658755 Chr.6: 151349313-151349313 on Build GRCh38
rs755383862 Chr.6: 151349300-151349300 on Build GRCh38
rs752192182 Chr.6: 151349489-151349489 on Build GRCh38
rs556828587 Chr.6: 151349279-151349279 on Build GRCh38
rs948721456 Chr.6: 151349352-151349352 on Build GRCh38
rs376468313 Chr.6: 151349187-151349187 on Build GRCh38
rs377106873 Chr.6: 151349197-151349199 on Build GRCh38
rs775211883 Chr.6: 151349160-151349160 on Build GRCh38
rs1016165791 Chr.6: 151349497-151349497 on Build GRCh38
rs141729078 Chr.6: 151349314-151349314 on Build GRCh38
rs142810400 Chr.6: 151349521-151349521 on Build GRCh38
rs142077552 Chr.6: 151349537-151349537 on Build GRCh38
rs748805861 Chr.6: 151349258-151349258 on Build GRCh38
rs111653642 Chr.6: 151349197-151349197 on Build GRCh38
rs761561267 Chr.6: 151349183-151349183 on Build GRCh38
rs377245112 Chr.6: 151349342-151349342 on Build GRCh38
rs979983116 Chr.6: 151349368-151349368 on Build GRCh38

View Full Product Details