Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 17333827-17334331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHR
Entrez Gene ID
196
Gene Name
aryl hydrocarbon receptor
Gene Aliases
bHLHe76
UniGene
Hs.171189
Gene Chromosome Location
Chr.7: 17298652-17346151 on Build GRCh38
Transcripts
NM_001621.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157301 c.811C>G p.P271A Chr.7: 17334017-17334017 on Build GRCh38
3778302 c.817C>A p.Q273K Chr.7: 17334023-17334023 on Build GRCh38
6597054 c.900T>C p.C300C Chr.7: 17334106-17334106 on Build GRCh38
7231083 c.772A>C p.I258L Chr.7: 17333978-17333978 on Build GRCh38
6451804 c.842G>A p.R281Q Chr.7: 17334048-17334048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374545462 Chr.7: 17334085-17334085 on Build GRCh38
rs547764617 Chr.7: 17334248-17334248 on Build GRCh38
rs759478631 Chr.7: 17333935-17333935 on Build GRCh38
rs978580509 Chr.7: 17334052-17334052 on Build GRCh38
rs960008625 Chr.7: 17334232-17334232 on Build GRCh38
rs550490159 Chr.7: 17333852-17333852 on Build GRCh38
rs776119669 Chr.7: 17334025-17334025 on Build GRCh38
rs750482862 Chr.7: 17333959-17333959 on Build GRCh38
rs778210325 Chr.7: 17334138-17334138 on Build GRCh38
rs369393068 Chr.7: 17334272-17334272 on Build GRCh38
rs2074113 Chr.7: 17334147-17334147 on Build GRCh38
rs746572500 Chr.7: 17334140-17334140 on Build GRCh38
rs969636661 Chr.7: 17333857-17333857 on Build GRCh38
rs760967749 Chr.7: 17334084-17334084 on Build GRCh38
rs140380733 Chr.7: 17334047-17334047 on Build GRCh38
rs752519254 Chr.7: 17333946-17333946 on Build GRCh38
rs775471702 Chr.7: 17334160-17334160 on Build GRCh38
rs942879053 Chr.7: 17333968-17333968 on Build GRCh38
rs767773568 Chr.7: 17334048-17334048 on Build GRCh38
rs922655270 Chr.7: 17334218-17334218 on Build GRCh38
rs772053459 Chr.7: 17334277-17334277 on Build GRCh38
rs527694635 Chr.7: 17334234-17334234 on Build GRCh38
rs148672036 Chr.7: 17333950-17333950 on Build GRCh38
rs777334061 Chr.7: 17334013-17334013 on Build GRCh38
rs750670730 Chr.7: 17334049-17334049 on Build GRCh38
rs764940539 Chr.7: 17333938-17333938 on Build GRCh38
rs771736391 Chr.7: 17334007-17334007 on Build GRCh38
rs142158496 Chr.7: 17333977-17333977 on Build GRCh38
rs779385734 Chr.7: 17333978-17333978 on Build GRCh38
rs772419357 Chr.7: 17334136-17334137 on Build GRCh38
rs1041022999 Chr.7: 17334192-17334192 on Build GRCh38
rs747737461 Chr.7: 17333892-17333892 on Build GRCh38
rs368233078 Chr.7: 17333872-17333872 on Build GRCh38
rs768991013 Chr.7: 17334247-17334247 on Build GRCh38
rs772733168 Chr.7: 17333899-17333899 on Build GRCh38
rs770894231 Chr.7: 17333919-17333919 on Build GRCh38
rs753076716 Chr.7: 17334133-17334133 on Build GRCh38
rs201235493 Chr.7: 17334298-17334298 on Build GRCh38
rs778604526 Chr.7: 17333885-17333886 on Build GRCh38
rs769687419 Chr.7: 17334154-17334154 on Build GRCh38
rs773495408 Chr.7: 17334141-17334142 on Build GRCh38
rs113673253 Chr.7: 17334030-17334030 on Build GRCh38
rs905661902 Chr.7: 17334185-17334185 on Build GRCh38
rs770286482 Chr.7: 17334019-17334019 on Build GRCh38
rs186541119 Chr.7: 17334142-17334142 on Build GRCh38
rs756620867 Chr.7: 17333871-17333871 on Build GRCh38
rs1035394113 Chr.7: 17333932-17333932 on Build GRCh38
rs376892230 Chr.7: 17334146-17334146 on Build GRCh38
rs562661363 Chr.7: 17333854-17333854 on Build GRCh38
rs779434112 Chr.7: 17333882-17333882 on Build GRCh38
rs748487723 Chr.7: 17333986-17333986 on Build GRCh38
rs566146872 Chr.7: 17334035-17334035 on Build GRCh38
rs771658060 Chr.7: 17333898-17333898 on Build GRCh38
rs760154499 Chr.7: 17333909-17333909 on Build GRCh38
rs748038586 Chr.7: 17334246-17334246 on Build GRCh38
rs769201930 Chr.7: 17333881-17333881 on Build GRCh38
rs911350740 Chr.7: 17333955-17333955 on Build GRCh38
rs755496764 Chr.7: 17334098-17334098 on Build GRCh38
rs758744308 Chr.7: 17334137-17334137 on Build GRCh38
rs780694305 Chr.7: 17333965-17333965 on Build GRCh38
rs973131402 Chr.7: 17334261-17334261 on Build GRCh38
rs375657110 Chr.7: 17333869-17333869 on Build GRCh38
rs1005684703 Chr.7: 17334263-17334263 on Build GRCh38
rs754340551 Chr.7: 17333971-17333971 on Build GRCh38
rs1025639342 Chr.7: 17334291-17334291 on Build GRCh38
rs765898500 Chr.7: 17334114-17334114 on Build GRCh38
rs113274268 Chr.7: 17334233-17334233 on Build GRCh38
rs754003410 Chr.7: 17334088-17334088 on Build GRCh38
rs1041663769 Chr.7: 17333970-17333970 on Build GRCh38
rs746512401 Chr.7: 17334014-17334014 on Build GRCh38
rs781291604 Chr.7: 17333868-17333868 on Build GRCh38
rs749667770 Chr.7: 17333878-17333878 on Build GRCh38
rs201499552 Chr.7: 17334212-17334216 on Build GRCh38
rs747810579 Chr.7: 17333886-17333886 on Build GRCh38
rs888540193 Chr.7: 17334253-17334253 on Build GRCh38
rs757735788 Chr.7: 17334176-17334177 on Build GRCh38

View Full Product Details