Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30921348-30921862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP1
Entrez Gene ID
358
Gene Name
aquaporin 1 (Colton blood group)
Gene Aliases
AQP-CHIP, CHIP28, CO
UniGene
Hs.76152
Gene Chromosome Location
Chr.7: 30911800-30925516 on Build GRCh38
Transcripts
NM_001185060.1 NM_001185061.1 NM_001185062.1 NM_198098.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
236996 c.135+2T>C p.? Chr.7: 30921840-30921840 on Build GRCh38
6347983 c.60G>T p.M20I Chr.7: 30921763-30921763 on Build GRCh38
4534971 c.117G>T p.V39V Chr.7: 30921820-30921820 on Build GRCh38
6548127 c.3G>T p.M1I Chr.7: 30921706-30921706 on Build GRCh38
3663153 c.75G>A p.K25K Chr.7: 30921778-30921778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556967160 Chr.7: 30921835-30921835 on Build GRCh38
rs997877079 Chr.7: 30921814-30921814 on Build GRCh38
rs1018430650 Chr.7: 30921642-30921642 on Build GRCh38
rs954915225 Chr.7: 30921732-30921734 on Build GRCh38
rs776813614 Chr.7: 30921685-30921685 on Build GRCh38
rs149264909 Chr.7: 30921568-30921568 on Build GRCh38
rs868506968 Chr.7: 30921715-30921715 on Build GRCh38
rs976937684 Chr.7: 30921517-30921517 on Build GRCh38
rs893464317 Chr.7: 30921633-30921633 on Build GRCh38
rs1005168781 Chr.7: 30921496-30921496 on Build GRCh38
rs1031109348 Chr.7: 30921441-30921441 on Build GRCh38
rs563326561 Chr.7: 30921447-30921447 on Build GRCh38
rs538254989 Chr.7: 30921763-30921763 on Build GRCh38
rs561208577 Chr.7: 30921477-30921477 on Build GRCh38
rs946649179 Chr.7: 30921577-30921577 on Build GRCh38
rs745336730 Chr.7: 30921558-30921558 on Build GRCh38
rs1042094942 Chr.7: 30921371-30921371 on Build GRCh38
rs974939860 Chr.7: 30921498-30921498 on Build GRCh38
rs764683412 Chr.7: 30921383-30921383 on Build GRCh38
rs773657460 Chr.7: 30921641-30921641 on Build GRCh38
rs776497596 Chr.7: 30921716-30921716 on Build GRCh38
rs528411526 Chr.7: 30921485-30921485 on Build GRCh38
rs2267720 Chr.7: 30921380-30921380 on Build GRCh38
rs1018395748 Chr.7: 30921506-30921506 on Build GRCh38
rs187798109 Chr.7: 30921717-30921717 on Build GRCh38
rs192763050 Chr.7: 30921627-30921627 on Build GRCh38
rs965253133 Chr.7: 30921495-30921495 on Build GRCh38
rs970653755 Chr.7: 30921436-30921436 on Build GRCh38
rs551946007 Chr.7: 30921625-30921625 on Build GRCh38
rs537605954 Chr.7: 30921635-30921635 on Build GRCh38
rs28362729 Chr.7: 30921708-30921708 on Build GRCh38
rs906441843 Chr.7: 30921622-30921622 on Build GRCh38
rs895945979 Chr.7: 30921384-30921384 on Build GRCh38
rs942105109 Chr.7: 30921548-30921548 on Build GRCh38
rs187858042 Chr.7: 30921472-30921472 on Build GRCh38
rs1024856832 Chr.7: 30921435-30921435 on Build GRCh38
rs750322155 Chr.7: 30921823-30921823 on Build GRCh38
rs963810261 Chr.7: 30921512-30921512 on Build GRCh38
rs1057457492 Chr.7: 30921648-30921648 on Build GRCh38
rs947883724 Chr.7: 30921587-30921587 on Build GRCh38
rs956450047 Chr.7: 30921827-30921827 on Build GRCh38
rs552531843 Chr.7: 30921714-30921714 on Build GRCh38
rs559811893 Chr.7: 30921532-30921532 on Build GRCh38
rs999144551 Chr.7: 30921438-30921438 on Build GRCh38
rs770421669 Chr.7: 30921712-30921712 on Build GRCh38
rs963825730 Chr.7: 30921684-30921684 on Build GRCh38
rs144480541 Chr.7: 30921711-30921711 on Build GRCh38
rs757255955 Chr.7: 30921781-30921781 on Build GRCh38
rs575352844 Chr.7: 30921471-30921471 on Build GRCh38
rs549337021 Chr.7: 30921647-30921647 on Build GRCh38
rs759512846 Chr.7: 30921700-30921700 on Build GRCh38
rs184128868 Chr.7: 30921656-30921656 on Build GRCh38
rs867023718 Chr.7: 30921593-30921593 on Build GRCh38
rs547796022 Chr.7: 30921520-30921520 on Build GRCh38
rs772573296 Chr.7: 30921634-30921634 on Build GRCh38
rs987963614 Chr.7: 30921829-30921829 on Build GRCh38
rs907832306 Chr.7: 30921817-30921817 on Build GRCh38
rs775299722 Chr.7: 30921620-30921620 on Build GRCh38
rs984130038 Chr.7: 30921783-30921783 on Build GRCh38
rs894992218 Chr.7: 30921393-30921393 on Build GRCh38
rs750771143 Chr.7: 30921672-30921672 on Build GRCh38
rs994263324 Chr.7: 30921704-30921704 on Build GRCh38
rs563058618 Chr.7: 30921399-30921399 on Build GRCh38
rs752919779 Chr.7: 30921710-30921710 on Build GRCh38
rs905946165 Chr.7: 30921377-30921377 on Build GRCh38
rs1012082384 Chr.7: 30921403-30921403 on Build GRCh38
rs964020546 Chr.7: 30921752-30921752 on Build GRCh38
rs780966313 Chr.7: 30921821-30921821 on Build GRCh38

View Full Product Details