Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30923343-30923849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP1
Entrez Gene ID
358
Gene Name
aquaporin 1 (Colton blood group)
Gene Aliases
AQP-CHIP, CHIP28, CO
UniGene
Hs.76152
Gene Chromosome Location
Chr.7: 30911800-30925516 on Build GRCh38
Transcripts
NM_001185060.1 NM_001185061.1 NM_001185062.1 NM_198098.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622781 c.779T>A p.I260N Chr.7: 30923598-30923598 on Build GRCh38
4402199 c.721C>T p.R241C Chr.7: 30923540-30923540 on Build GRCh38
6110063 c.748G>A p.V250M Chr.7: 30923567-30923567 on Build GRCh38
3637960 c.522C>T p.A174A Chr.7: 30923590-30923590 on Build GRCh38
297655 c.675C>T p.L225L Chr.7: 30923494-30923494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777591464 Chr.7: 30923811-30923811 on Build GRCh38
rs752486311 Chr.7: 30923799-30923799 on Build GRCh38
rs931917879 Chr.7: 30923539-30923539 on Build GRCh38
rs28362739 Chr.7: 30923694-30923695 on Build GRCh38
rs755861040 Chr.7: 30923795-30923798 on Build GRCh38
rs1049739937 Chr.7: 30923542-30923542 on Build GRCh38
rs770424088 Chr.7: 30923644-30923644 on Build GRCh38
rs907623223 Chr.7: 30923691-30923691 on Build GRCh38
rs779725060 Chr.7: 30923394-30923394 on Build GRCh38
rs760999882 Chr.7: 30923591-30923591 on Build GRCh38
rs901819120 Chr.7: 30923681-30923681 on Build GRCh38
rs752063738 Chr.7: 30923474-30923474 on Build GRCh38
rs759697092 Chr.7: 30923784-30923784 on Build GRCh38
rs911812863 Chr.7: 30923793-30923793 on Build GRCh38
rs979660393 Chr.7: 30923821-30923821 on Build GRCh38
rs772541900 Chr.7: 30923551-30923551 on Build GRCh38
rs766446681 Chr.7: 30923678-30923678 on Build GRCh38
rs762347784 Chr.7: 30923583-30923583 on Build GRCh38
rs1026363459 Chr.7: 30923690-30923690 on Build GRCh38
rs963138473 Chr.7: 30923715-30923715 on Build GRCh38
rs973245291 Chr.7: 30923752-30923752 on Build GRCh38
rs767634051 Chr.7: 30923480-30923480 on Build GRCh38
rs746735108 Chr.7: 30923553-30923553 on Build GRCh38
rs765650073 Chr.7: 30923463-30923463 on Build GRCh38
rs988985986 Chr.7: 30923780-30923780 on Build GRCh38
rs752645594 Chr.7: 30923631-30923631 on Build GRCh38
rs943984109 Chr.7: 30923794-30923794 on Build GRCh38
rs180936597 Chr.7: 30923807-30923807 on Build GRCh38
rs914070676 Chr.7: 30923489-30923489 on Build GRCh38
rs140536387 Chr.7: 30923674-30923674 on Build GRCh38
rs149637560 Chr.7: 30923561-30923561 on Build GRCh38
rs375975852 Chr.7: 30923473-30923473 on Build GRCh38
rs1052094212 Chr.7: 30923778-30923778 on Build GRCh38
rs376146000 Chr.7: 30923424-30923424 on Build GRCh38
rs780739432 Chr.7: 30923647-30923647 on Build GRCh38
rs575472927 Chr.7: 30923587-30923587 on Build GRCh38
rs748039521 Chr.7: 30923540-30923540 on Build GRCh38
rs757443094 Chr.7: 30923476-30923476 on Build GRCh38
rs966843251 Chr.7: 30923386-30923386 on Build GRCh38
rs1021084299 Chr.7: 30923790-30923790 on Build GRCh38
rs760695211 Chr.7: 30923675-30923675 on Build GRCh38
rs781587140 Chr.7: 30923405-30923405 on Build GRCh38
rs746033215 Chr.7: 30923414-30923414 on Build GRCh38
rs1021064241 Chr.7: 30923374-30923374 on Build GRCh38
rs375226621 Chr.7: 30923520-30923520 on Build GRCh38
rs754801843 Chr.7: 30923598-30923598 on Build GRCh38
rs758320085 Chr.7: 30923635-30923635 on Build GRCh38
rs375061904 Chr.7: 30923593-30923593 on Build GRCh38
rs144238098 Chr.7: 30923577-30923577 on Build GRCh38
rs201414781 Chr.7: 30923566-30923566 on Build GRCh38
rs745338048 Chr.7: 30923791-30923792 on Build GRCh38
rs775705720 Chr.7: 30923467-30923467 on Build GRCh38
rs554574639 Chr.7: 30923673-30923673 on Build GRCh38
rs764513622 Chr.7: 30923575-30923575 on Build GRCh38
rs755937705 Chr.7: 30923423-30923423 on Build GRCh38
rs566538624 Chr.7: 30923434-30923434 on Build GRCh38
rs777409509 Chr.7: 30923639-30923639 on Build GRCh38
rs1035349679 Chr.7: 30923661-30923661 on Build GRCh38
rs761409742 Chr.7: 30923594-30923594 on Build GRCh38
rs769152616 Chr.7: 30923654-30923654 on Build GRCh38
rs1026850424 Chr.7: 30923629-30923629 on Build GRCh38
rs765076430 Chr.7: 30923798-30923798 on Build GRCh38
rs563208076 Chr.7: 30923365-30923365 on Build GRCh38
rs1001031938 Chr.7: 30923597-30923597 on Build GRCh38
rs905828486 Chr.7: 30923652-30923652 on Build GRCh38
rs746686966 Chr.7: 30923411-30923411 on Build GRCh38
rs950786558 Chr.7: 30923695-30923695 on Build GRCh38
rs779968706 Chr.7: 30923515-30923515 on Build GRCh38
rs556135632 Chr.7: 30923643-30923643 on Build GRCh38
rs1025237855 Chr.7: 30923658-30923658 on Build GRCh38
rs749302762 Chr.7: 30923428-30923428 on Build GRCh38
rs151294803 Chr.7: 30923432-30923432 on Build GRCh38
rs997536693 Chr.7: 30923685-30923685 on Build GRCh38
rs140239885 Chr.7: 30923500-30923500 on Build GRCh38
rs189379708 Chr.7: 30923392-30923392 on Build GRCh38
rs571614381 Chr.7: 30923681-30923682 on Build GRCh38
rs67184369 Chr.7: 30923684-30923685 on Build GRCh38
rs749008815 Chr.7: 30923519-30923519 on Build GRCh38
rs969702447 Chr.7: 30923395-30923395 on Build GRCh38
rs77695754 Chr.7: 30923439-30923439 on Build GRCh38
rs199860979 Chr.7: 30923560-30923560 on Build GRCh38
rs796517133 Chr.7: 30923693-30923694 on Build GRCh38
rs200837934 Chr.7: 30923653-30923653 on Build GRCh38
rs536215414 Chr.7: 30923669-30923669 on Build GRCh38
rs921791755 Chr.7: 30923756-30923756 on Build GRCh38
rs778613724 Chr.7: 30923620-30923620 on Build GRCh38
rs931848912 Chr.7: 30923759-30923759 on Build GRCh38
rs779654022 Chr.7: 30923425-30923425 on Build GRCh38
rs760178951 Chr.7: 30923461-30923461 on Build GRCh38
rs200924730 Chr.7: 30923438-30923438 on Build GRCh38
rs983133460 Chr.7: 30923458-30923458 on Build GRCh38
rs750461875 Chr.7: 30923590-30923590 on Build GRCh38
rs386711778 Chr.7: 30923677-30923678 on Build GRCh38
rs932635477 Chr.7: 30923534-30923534 on Build GRCh38
rs995400360 Chr.7: 30923623-30923623 on Build GRCh38
rs970329460 Chr.7: 30923662-30923662 on Build GRCh38
rs771575765 Chr.7: 30923543-30923543 on Build GRCh38
rs750670054 Chr.7: 30923484-30923484 on Build GRCh38

View Full Product Details