Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37848464-37849004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

SNP Detail

SNP ID Chromosome Location
rs1049955855 Chr.7: 37848823-37848823 on Build GRCh38
rs991045654 Chr.7: 37848729-37848729 on Build GRCh38
rs116102937 Chr.7: 37848529-37848529 on Build GRCh38
rs1025904868 Chr.7: 37848790-37848790 on Build GRCh38
rs898625545 Chr.7: 37848946-37848946 on Build GRCh38
rs898678643 Chr.7: 37848782-37848782 on Build GRCh38
rs915029963 Chr.7: 37848875-37848875 on Build GRCh38
rs2598048 Chr.7: 37848572-37848572 on Build GRCh38
rs992789320 Chr.7: 37848648-37848648 on Build GRCh38
rs1051185358 Chr.7: 37848892-37848892 on Build GRCh38
rs80144288 Chr.7: 37848758-37848758 on Build GRCh38
rs35922143 Chr.7: 37848540-37848540 on Build GRCh38
rs571881008 Chr.7: 37848956-37848956 on Build GRCh38
rs754583940 Chr.7: 37848856-37848856 on Build GRCh38
rs76138543 Chr.7: 37848586-37848586 on Build GRCh38
rs115132982 Chr.7: 37848880-37848880 on Build GRCh38
rs377655054 Chr.7: 37848754-37848754 on Build GRCh38
rs889522911 Chr.7: 37848762-37848762 on Build GRCh38
rs921397 Chr.7: 37848972-37848972 on Build GRCh38
rs545249773 Chr.7: 37848960-37848960 on Build GRCh38
rs183600800 Chr.7: 37848646-37848646 on Build GRCh38
rs933940138 Chr.7: 37848890-37848890 on Build GRCh38
rs2598047 Chr.7: 37848886-37848886 on Build GRCh38
rs35876675 Chr.7: 37848503-37848503 on Build GRCh38
rs1042295820 Chr.7: 37848745-37848745 on Build GRCh38
rs1017499085 Chr.7: 37848776-37848776 on Build GRCh38
rs762715298 Chr.7: 37848602-37848602 on Build GRCh38
rs946612452 Chr.7: 37848740-37848740 on Build GRCh38
rs759594717 Chr.7: 37848709-37848709 on Build GRCh38
rs374264712 Chr.7: 37848785-37848785 on Build GRCh38
rs559376205 Chr.7: 37848884-37848884 on Build GRCh38
rs758787380 Chr.7: 37848917-37848917 on Build GRCh38
rs1031733224 Chr.7: 37848643-37848643 on Build GRCh38
rs4723713 Chr.7: 37848752-37848752 on Build GRCh38
rs1027547601 Chr.7: 37848967-37848967 on Build GRCh38
rs971680816 Chr.7: 37848798-37848798 on Build GRCh38
rs185352152 Chr.7: 37848539-37848539 on Build GRCh38
rs932880025 Chr.7: 37848807-37848807 on Build GRCh38
rs577710747 Chr.7: 37848913-37848913 on Build GRCh38
rs927368654 Chr.7: 37848655-37848655 on Build GRCh38
rs1028908867 Chr.7: 37848574-37848574 on Build GRCh38
rs77025462 Chr.7: 37848552-37848552 on Build GRCh38
rs1017133925 Chr.7: 37848553-37848553 on Build GRCh38
rs971396255 Chr.7: 37848642-37848642 on Build GRCh38
rs190608230 Chr.7: 37848607-37848607 on Build GRCh38
rs990269045 Chr.7: 37848871-37848871 on Build GRCh38
rs1038131788 Chr.7: 37848929-37848929 on Build GRCh38
rs979982759 Chr.7: 37848716-37848716 on Build GRCh38
rs1006665711 Chr.7: 37848765-37848765 on Build GRCh38
rs963272961 Chr.7: 37848562-37848562 on Build GRCh38
rs757640180 Chr.7: 37848939-37848939 on Build GRCh38
rs16879722 Chr.7: 37848516-37848516 on Build GRCh38
rs915103144 Chr.7: 37848739-37848739 on Build GRCh38
rs529217643 Chr.7: 37848810-37848810 on Build GRCh38
rs551942725 Chr.7: 37848916-37848916 on Build GRCh38
rs901385086 Chr.7: 37848566-37848566 on Build GRCh38
rs1034382047 Chr.7: 37848833-37848833 on Build GRCh38
rs1000269984 Chr.7: 37848600-37848600 on Build GRCh38
rs970376736 Chr.7: 37848662-37848662 on Build GRCh38
rs534850205 Chr.7: 37848491-37848491 on Build GRCh38
rs373433456 Chr.7: 37848656-37848656 on Build GRCh38
rs994387581 Chr.7: 37848789-37848789 on Build GRCh38

View Full Product Details