Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38383573-38384084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
UniGene
Hs.592182
Gene Chromosome Location
Chr.7: 38383694-38631567 on Build GRCh38
Transcripts
NM_001635.3 NM_139316.2 XM_006715689.3 XM_006715690.3 XM_011515271.2 XM_017011995.1 XM_017011996.1

SNP Detail

SNP ID Chromosome Location
rs998809941 Chr.7: 38384020-38384020 on Build GRCh38
rs138178389 Chr.7: 38383880-38383880 on Build GRCh38
rs1054589992 Chr.7: 38383721-38383721 on Build GRCh38
rs1041216430 Chr.7: 38383992-38383992 on Build GRCh38
rs770781806 Chr.7: 38384042-38384042 on Build GRCh38
rs185153914 Chr.7: 38383626-38383626 on Build GRCh38
rs943790347 Chr.7: 38383930-38383930 on Build GRCh38
rs929568125 Chr.7: 38383862-38383862 on Build GRCh38
rs540640481 Chr.7: 38384016-38384016 on Build GRCh38
rs949764710 Chr.7: 38383606-38383606 on Build GRCh38
rs372955983 Chr.7: 38383829-38383829 on Build GRCh38
rs1006720566 Chr.7: 38384014-38384014 on Build GRCh38
rs972379350 Chr.7: 38383597-38383597 on Build GRCh38
rs397740441 Chr.7: 38383687-38383687 on Build GRCh38
rs189814356 Chr.7: 38383649-38383649 on Build GRCh38
rs899604870 Chr.7: 38384005-38384005 on Build GRCh38
rs918269670 Chr.7: 38383599-38383599 on Build GRCh38
rs904404036 Chr.7: 38384015-38384015 on Build GRCh38
rs1029955424 Chr.7: 38383998-38383998 on Build GRCh38
rs549402256 Chr.7: 38383777-38383777 on Build GRCh38
rs370684770 Chr.7: 38383670-38383671 on Build GRCh38
rs556330220 Chr.7: 38383614-38383614 on Build GRCh38
rs572121093 Chr.7: 38383906-38383906 on Build GRCh38
rs889749291 Chr.7: 38384007-38384007 on Build GRCh38
rs35466776 Chr.7: 38383671-38383672 on Build GRCh38
rs893211727 Chr.7: 38383723-38383723 on Build GRCh38
rs972452701 Chr.7: 38384048-38384048 on Build GRCh38
rs1016805298 Chr.7: 38384041-38384041 on Build GRCh38
rs983023056 Chr.7: 38383661-38383661 on Build GRCh38
rs920815460 Chr.7: 38383852-38383852 on Build GRCh38
rs997535532 Chr.7: 38383991-38383991 on Build GRCh38
rs1017153178 Chr.7: 38383710-38383710 on Build GRCh38
rs77661374 Chr.7: 38383671-38383671 on Build GRCh38
rs35923445 Chr.7: 38383687-38383688 on Build GRCh38
rs1041804281 Chr.7: 38383837-38383837 on Build GRCh38

View Full Product Details