The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242648.2 | NP_001229577.1 | 3 - 4 | 438 | 109 |
NM_001322308.1 | NP_001309237.1 | 2 - 3 | 393 | 109 | |
NM_001322309.1 | NP_001309238.1 | 2 - 3 | 284 | 109 | |
NM_001322310.1 | NP_001309239.1 | 1 - 2 | 152 | 109 | |
NM_030948.3 | NP_112210.1 | 3 - 4 | 628 | 109 | |
XM_005248934.2 | XP_005248991.1 | 3 - 4 | 470 | 109 | |
XM_017010454.1 | XP_016865943.1 | 3 - 4 | 470 | 109 | |
XM_017010455.1 | XP_016865944.1 | 3 - 4 | 438 | 109 | |
XM_017010456.1 | XP_016865945.1 | 2 - 3 | 435 | 109 | |
XM_017010457.1 | XP_016865946.1 | 2 - 3 | 414 | 109 | |
XM_017010459.1 | XP_016865948.1 | 3 - 4 | 470 | 109 | |
XM_017010460.1 | XP_016865949.1 | 3 - 4 | 438 | 109 | |
XM_017010461.1 | XP_016865950.1 | 2 - 3 | 409 | 109 | |
XM_017010462.1 | XP_016865951.1 | 2 - 3 | 435 | 109 | |
GenBank mRNA | AB051520.1 | - | 2 - 3 | 235 | 109 |
AK090769.1 | - | 2 - 3 | 295 | 109 | |
AK290017.1 | - | 3 - 4 | 289 | 109 | |
AK295263.1 | - | 3 - 4 | 438 | 109 | |
BC047159.1 | - | 2 - 3 | 283 | 109 | |
BC101526.1 | - | 2 - 3 | 202 | 109 | |
BC101528.1 | - | 2 - 3 | 202 | 109 | |
CR746945.1 | - | 2 - 3 | 294 | 109 | |
DA253448.1 | - | 3 - 4 | 470 | 109 | |
DA330657.1 | - | 3 - 4 | 289 | 109 |