The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080433.1 | NP_001073902.1 | 2 - 3 | 1744 | 68 |
XM_005264125.1 | XP_005264182.1 | 2 - 3 | 1269 | 68 | |
GenBank mRNA | AB067499.2 | - | 2 - 3 | 1828 | 68 |
BC131557.1 | - | 2 - 3 | 416 | 68 |