Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66628695-66629213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391919 c.86A>T p.E29V Chr.7: 66629150-66629150 on Build GRCh38
6408632 c.50C>T p.A17V Chr.7: 66629114-66629114 on Build GRCh38
7314708 c.89_100del12 p.P33_T36delPTAT Chr.7: 66629153-66629164 on Build GRCh38
6988019 c.35G>T p.R12L Chr.7: 66629099-66629099 on Build GRCh38
6343921 c.121G>T p.A41S Chr.7: 66629185-66629185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777567549 Chr.7: 66629017-66629017 on Build GRCh38
rs1037956745 Chr.7: 66628810-66628810 on Build GRCh38
rs886062413 Chr.7: 66629082-66629082 on Build GRCh38
rs933610815 Chr.7: 66629076-66629076 on Build GRCh38
rs1013892362 Chr.7: 66628735-66628735 on Build GRCh38
rs763086570 Chr.7: 66629180-66629180 on Build GRCh38
rs923977363 Chr.7: 66628751-66628751 on Build GRCh38
rs761974240 Chr.7: 66629159-66629159 on Build GRCh38
rs766882316 Chr.7: 66629101-66629101 on Build GRCh38
rs1013070683 Chr.7: 66628725-66628725 on Build GRCh38
rs545156204 Chr.7: 66628782-66628782 on Build GRCh38
rs996330325 Chr.7: 66628829-66628829 on Build GRCh38
rs982415840 Chr.7: 66628900-66628900 on Build GRCh38
rs373655601 Chr.7: 66629177-66629177 on Build GRCh38
rs886062411 Chr.7: 66628978-66628978 on Build GRCh38
rs958093696 Chr.7: 66628982-66628982 on Build GRCh38
rs765260614 Chr.7: 66629096-66629096 on Build GRCh38
rs940237483 Chr.7: 66629012-66629012 on Build GRCh38
rs1052412405 Chr.7: 66628968-66628968 on Build GRCh38
rs751937729 Chr.7: 66629109-66629109 on Build GRCh38
rs530783681 Chr.7: 66628787-66628787 on Build GRCh38
rs989398645 Chr.7: 66628753-66628753 on Build GRCh38
rs557231998 Chr.7: 66628969-66628969 on Build GRCh38
rs778647590 Chr.7: 66629165-66629176 on Build GRCh38
rs760486366 Chr.7: 66629043-66629043 on Build GRCh38
rs886062409 Chr.7: 66628920-66628920 on Build GRCh38
rs754475798 Chr.7: 66629163-66629174 on Build GRCh38
rs764365979 Chr.7: 66629059-66629059 on Build GRCh38
rs929767135 Chr.7: 66628910-66628910 on Build GRCh38
rs371919994 Chr.7: 66629140-66629140 on Build GRCh38
rs927833835 Chr.7: 66628904-66628904 on Build GRCh38
rs777578179 Chr.7: 66629117-66629117 on Build GRCh38
rs1039397893 Chr.7: 66629044-66629044 on Build GRCh38
rs756855275 Chr.7: 66629127-66629127 on Build GRCh38
rs1010933920 Chr.7: 66628993-66628993 on Build GRCh38
rs947900907 Chr.7: 66628772-66628772 on Build GRCh38
rs947263110 Chr.7: 66628842-66628842 on Build GRCh38
rs886062407 Chr.7: 66628884-66628884 on Build GRCh38
rs753399965 Chr.7: 66629153-66629164 on Build GRCh38
rs574026019 Chr.7: 66628995-66628995 on Build GRCh38
rs955013476 Chr.7: 66628838-66628838 on Build GRCh38
rs771944011 Chr.7: 66629137-66629137 on Build GRCh38
rs750951765 Chr.7: 66629098-66629098 on Build GRCh38
rs1031117251 Chr.7: 66628975-66628975 on Build GRCh38
rs996454212 Chr.7: 66628965-66628965 on Build GRCh38
rs529283909 Chr.7: 66628848-66628848 on Build GRCh38
rs954805706 Chr.7: 66628749-66628749 on Build GRCh38
rs551906037 Chr.7: 66628949-66628949 on Build GRCh38
rs760746593 Chr.7: 66629031-66629032 on Build GRCh38
rs1020470706 Chr.7: 66628915-66628915 on Build GRCh38
rs985987241 Chr.7: 66628836-66628836 on Build GRCh38
rs868184670 Chr.7: 66629124-66629124 on Build GRCh38
rs1004835198 Chr.7: 66628897-66628897 on Build GRCh38
rs958062363 Chr.7: 66628867-66628867 on Build GRCh38
rs886062412 Chr.7: 66629019-66629019 on Build GRCh38
rs1000099581 Chr.7: 66628972-66628972 on Build GRCh38
rs941097319 Chr.7: 66629024-66629024 on Build GRCh38
rs989472283 Chr.7: 66628873-66628873 on Build GRCh38
rs978557739 Chr.7: 66628750-66628750 on Build GRCh38
rs916495600 Chr.7: 66628766-66628766 on Build GRCh38
rs753471689 Chr.7: 66629125-66629125 on Build GRCh38
rs1015855393 Chr.7: 66628906-66628906 on Build GRCh38
rs1046817016 Chr.7: 66628946-66628946 on Build GRCh38
rs770506126 Chr.7: 66629022-66629022 on Build GRCh38
rs927832797 Chr.7: 66629010-66629010 on Build GRCh38
rs924567744 Chr.7: 66628876-66628877 on Build GRCh38
rs900755879 Chr.7: 66628817-66628817 on Build GRCh38
rs35526611 Chr.7: 66629021-66629021 on Build GRCh38
rs186270651 Chr.7: 66628785-66628785 on Build GRCh38
rs894611587 Chr.7: 66628717-66628717 on Build GRCh38
rs752044252 Chr.7: 66629192-66629192 on Build GRCh38
rs1007276581 Chr.7: 66629145-66629145 on Build GRCh38
rs868346951 Chr.7: 66628752-66628752 on Build GRCh38
rs374505432 Chr.7: 66629016-66629016 on Build GRCh38
rs1039332754 Chr.7: 66628958-66628958 on Build GRCh38
rs191169882 Chr.7: 66628895-66628895 on Build GRCh38
rs567609628 Chr.7: 66628837-66628837 on Build GRCh38
rs897477257 Chr.7: 66628907-66628907 on Build GRCh38
rs774430442 Chr.7: 66629026-66629026 on Build GRCh38
rs755360166 Chr.7: 66629113-66629113 on Build GRCh38
rs571516346 Chr.7: 66628951-66628951 on Build GRCh38
rs920934895 Chr.7: 66629045-66629045 on Build GRCh38
rs769985127 Chr.7: 66629166-66629166 on Build GRCh38
rs534830374 Chr.7: 66628938-66628938 on Build GRCh38
rs982060715 Chr.7: 66629009-66629009 on Build GRCh38
rs747381612 Chr.7: 66629015-66629015 on Build GRCh38
rs888728946 Chr.7: 66628776-66628776 on Build GRCh38
rs536230559 Chr.7: 66628996-66628996 on Build GRCh38
rs916528162 Chr.7: 66628891-66628891 on Build GRCh38
rs1057134650 Chr.7: 66628886-66628886 on Build GRCh38
rs758648043 Chr.7: 66628952-66628952 on Build GRCh38
rs906300220 Chr.7: 66628781-66628781 on Build GRCh38
rs890695139 Chr.7: 66629153-66629153 on Build GRCh38
rs773209854 Chr.7: 66629171-66629171 on Build GRCh38
rs886062410 Chr.7: 66628971-66628971 on Build GRCh38
rs566129756 Chr.7: 66628903-66628903 on Build GRCh38
rs763423181 Chr.7: 66629099-66629099 on Build GRCh38
rs1046215393 Chr.7: 66628773-66628773 on Build GRCh38
rs796052690 Chr.7: 66629114-66629114 on Build GRCh38
rs761888221 Chr.7: 66629094-66629094 on Build GRCh38
rs139497775 Chr.7: 66628788-66628788 on Build GRCh38
rs1030069313 Chr.7: 66628922-66628922 on Build GRCh38
rs776777389 Chr.7: 66629061-66629061 on Build GRCh38
rs886062408 Chr.7: 66628910-66628910 on Build GRCh38
rs953135725 Chr.7: 66628929-66628929 on Build GRCh38
rs778513261 Chr.7: 66629130-66629130 on Build GRCh38
rs537743017 Chr.7: 66629011-66629011 on Build GRCh38
rs1007328828 Chr.7: 66628931-66628931 on Build GRCh38
rs745360140 Chr.7: 66629132-66629132 on Build GRCh38
rs908891854 Chr.7: 66628885-66628907 on Build GRCh38
rs766856368 Chr.7: 66629185-66629185 on Build GRCh38
rs776871737 Chr.7: 66629157-66629157 on Build GRCh38
rs766409684 Chr.7: 66629035-66629035 on Build GRCh38
rs768374078 Chr.7: 66629150-66629150 on Build GRCh38
rs537288495 Chr.7: 66628960-66628960 on Build GRCh38
rs1031464468 Chr.7: 66628862-66628862 on Build GRCh38
rs368529427 Chr.7: 66629033-66629033 on Build GRCh38
rs771922388 Chr.7: 66629037-66629037 on Build GRCh38

View Full Product Details