Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159808083-159808571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL6
Entrez Gene ID
343413
Gene Name
Fc receptor like 6
Gene Aliases
FcRH6
UniGene
Hs.196955
Gene Chromosome Location
Chr.1: 159800506-159816257 on Build GRCh38
Transcripts
NM_001004310.2 NM_001284217.1 XM_005245128.3 XM_005245129.3 XM_005245131.3 XM_006711292.3 XM_011509480.1 XM_011509481.2 XM_017001176.1 XM_017001177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5844137 c.233G>A p.R78H Chr.1: 159808358-159808358 on Build GRCh38
1165168 c.124T>A p.W42R Chr.1: 159808249-159808249 on Build GRCh38
6662960 c.163G>T p.D55Y Chr.1: 159808288-159808288 on Build GRCh38
2084707 c.259G>T p.V87L Chr.1: 159808384-159808384 on Build GRCh38
4684262 c.299C>T p.T100I Chr.1: 159808424-159808424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371528311 Chr.1: 159808312-159808312 on Build GRCh38
rs369432261 Chr.1: 159808304-159808304 on Build GRCh38
rs775504142 Chr.1: 159808381-159808381 on Build GRCh38
rs775555390 Chr.1: 159808285-159808285 on Build GRCh38
rs191439728 Chr.1: 159808524-159808524 on Build GRCh38
rs750374907 Chr.1: 159808331-159808331 on Build GRCh38
rs746257402 Chr.1: 159808470-159808470 on Build GRCh38
rs925142301 Chr.1: 159808346-159808346 on Build GRCh38
rs888640535 Chr.1: 159808486-159808486 on Build GRCh38
rs766872773 Chr.1: 159808507-159808507 on Build GRCh38
rs745393359 Chr.1: 159808180-159808180 on Build GRCh38
rs758761050 Chr.1: 159808537-159808537 on Build GRCh38
rs770158995 Chr.1: 159808485-159808485 on Build GRCh38
rs559398248 Chr.1: 159808498-159808498 on Build GRCh38
rs771757417 Chr.1: 159808199-159808199 on Build GRCh38
rs200331672 Chr.1: 159808449-159808449 on Build GRCh38
rs575326015 Chr.1: 159808440-159808440 on Build GRCh38
rs137931226 Chr.1: 159808400-159808400 on Build GRCh38
rs773614473 Chr.1: 159808490-159808490 on Build GRCh38
rs546662787 Chr.1: 159808143-159808143 on Build GRCh38
rs754561885 Chr.1: 159808354-159808354 on Build GRCh38
rs749462389 Chr.1: 159808540-159808540 on Build GRCh38
rs759701256 Chr.1: 159808157-159808157 on Build GRCh38
rs765409145 Chr.1: 159808191-159808191 on Build GRCh38
rs144777724 Chr.1: 159808430-159808430 on Build GRCh38
rs372542430 Chr.1: 159808447-159808447 on Build GRCh38
rs779211397 Chr.1: 159808458-159808458 on Build GRCh38
rs749590419 Chr.1: 159808267-159808267 on Build GRCh38
rs140964607 Chr.1: 159808339-159808339 on Build GRCh38
rs766397177 Chr.1: 159808418-159808418 on Build GRCh38
rs753754969 Chr.1: 159808174-159808174 on Build GRCh38
rs760823899 Chr.1: 159808239-159808239 on Build GRCh38
rs759530281 Chr.1: 159808500-159808500 on Build GRCh38
rs142135373 Chr.1: 159808526-159808526 on Build GRCh38
rs140069687 Chr.1: 159808375-159808375 on Build GRCh38
rs267598115 Chr.1: 159808245-159808245 on Build GRCh38
rs142957693 Chr.1: 159808365-159808365 on Build GRCh38
rs755417663 Chr.1: 159808320-159808320 on Build GRCh38
rs150017462 Chr.1: 159808325-159808325 on Build GRCh38
rs997385843 Chr.1: 159808229-159808229 on Build GRCh38
rs765562506 Chr.1: 159808131-159808131 on Build GRCh38
rs570234860 Chr.1: 159808206-159808206 on Build GRCh38
rs772602715 Chr.1: 159808380-159808380 on Build GRCh38
rs750427863 Chr.1: 159808248-159808248 on Build GRCh38
rs751670102 Chr.1: 159808156-159808156 on Build GRCh38
rs1052427646 Chr.1: 159808109-159808109 on Build GRCh38
rs776787242 Chr.1: 159808482-159808482 on Build GRCh38
rs779266111 Chr.1: 159808369-159808369 on Build GRCh38
rs748977146 Chr.1: 159808449-159808449 on Build GRCh38
rs373331988 Chr.1: 159808480-159808480 on Build GRCh38
rs544992174 Chr.1: 159808541-159808541 on Build GRCh38
rs758535382 Chr.1: 159808232-159808233 on Build GRCh38
rs542844278 Chr.1: 159808479-159808479 on Build GRCh38
rs751551791 Chr.1: 159808427-159808427 on Build GRCh38
rs780773565 Chr.1: 159808244-159808258 on Build GRCh38
rs751053507 Chr.1: 159808342-159808342 on Build GRCh38
rs375795262 Chr.1: 159808290-159808290 on Build GRCh38
rs961663335 Chr.1: 159808270-159808270 on Build GRCh38
rs754438434 Chr.1: 159808429-159808429 on Build GRCh38
rs185267666 Chr.1: 159808223-159808223 on Build GRCh38
rs771364171 Chr.1: 159808279-159808279 on Build GRCh38
rs756505701 Chr.1: 159808497-159808497 on Build GRCh38
rs760273352 Chr.1: 159808519-159808519 on Build GRCh38
rs757150227 Chr.1: 159808175-159808175 on Build GRCh38
rs765945746 Chr.1: 159808341-159808341 on Build GRCh38
rs758816105 Chr.1: 159808472-159808472 on Build GRCh38
rs752905425 Chr.1: 159808238-159808238 on Build GRCh38
rs190423836 Chr.1: 159808284-159808284 on Build GRCh38
rs372624549 Chr.1: 159808253-159808253 on Build GRCh38
rs762154232 Chr.1: 159808391-159808391 on Build GRCh38
rs778451111 Chr.1: 159808176-159808176 on Build GRCh38
rs116501788 Chr.1: 159808169-159808169 on Build GRCh38
rs552904423 Chr.1: 159808330-159808330 on Build GRCh38
rs747038435 Chr.1: 159808386-159808386 on Build GRCh38
rs750819528 Chr.1: 159808417-159808417 on Build GRCh38
rs750468964 Chr.1: 159808218-159808220 on Build GRCh38
rs572934645 Chr.1: 159808508-159808508 on Build GRCh38
rs768822377 Chr.1: 159808388-159808388 on Build GRCh38
rs762247602 Chr.1: 159808128-159808128 on Build GRCh38
rs537679761 Chr.1: 159808212-159808212 on Build GRCh38
rs567787373 Chr.1: 159808260-159808260 on Build GRCh38
rs562039899 Chr.1: 159808224-159808224 on Build GRCh38
rs944003944 Chr.1: 159808389-159808389 on Build GRCh38
rs147917646 Chr.1: 159808358-159808358 on Build GRCh38
rs1041499254 Chr.1: 159808217-159808217 on Build GRCh38
rs757170232 Chr.1: 159808246-159808246 on Build GRCh38
rs780674077 Chr.1: 159808357-159808357 on Build GRCh38
rs368990639 Chr.1: 159808233-159808233 on Build GRCh38
rs765055607 Chr.1: 159808393-159808393 on Build GRCh38
rs778973075 Chr.1: 159808247-159808247 on Build GRCh38
rs372255252 Chr.1: 159808173-159808173 on Build GRCh38
rs763885950 Chr.1: 159808240-159808240 on Build GRCh38
rs781360086 Chr.1: 159808356-159808356 on Build GRCh38
rs748338864 Chr.1: 159808484-159808484 on Build GRCh38
rs774831597 Chr.1: 159808282-159808282 on Build GRCh38
rs552115685 Chr.1: 159808200-159808200 on Build GRCh38
rs770182446 Chr.1: 159808445-159808445 on Build GRCh38
rs766414224 Chr.1: 159808144-159808144 on Build GRCh38
rs764249166 Chr.1: 159808296-159808296 on Build GRCh38
rs759578511 Chr.1: 159808237-159808237 on Build GRCh38
rs180697391 Chr.1: 159808136-159808136 on Build GRCh38
rs866572573 Chr.1: 159808315-159808315 on Build GRCh38
rs746591505 Chr.1: 159808259-159808259 on Build GRCh38
rs767700901 Chr.1: 159808161-159808161 on Build GRCh38
rs760091563 Chr.1: 159808283-159808283 on Build GRCh38
rs771359767 Chr.1: 159808232-159808232 on Build GRCh38
rs752080265 Chr.1: 159808258-159808259 on Build GRCh38

View Full Product Details