Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCLT1
Entrez Gene ID
132320
Gene Name
sodium channel and clathrin linker 1
Gene Aliases
CAP-1A, CAP1A
Chromosome Location
Chr.4: 128875206 - 129093607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144643.3 NP_653244.2 14 - 15 1723 114
XM_017007716.1 XP_016863205.1 14 - 15 1723 114
XM_017007717.1 XP_016863206.1 14 - 15 1723 114
XM_017007718.1 XP_016863207.1 14 - 15 1723 114
XM_017007719.1 XP_016863208.1 14 - 15 1244 114
XM_017007720.1 XP_016863209.1 15 - 16 1635 114
XM_017007721.1 XP_016863210.1 8 - 9 837 114
GenBank mRNA AK055217.1 - 14 - 15 1723 114
AK122852.1 - 4 - 5 2346 114

View Full Product Details