The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018704.2 | NP_061174.1 | 6 - 6 | 3663 | 67 |
XM_011541781.2 | XP_011540083.1 | 6 - 6 | 3734 | 67 | |
XM_017001806.1 | XP_016857295.1 | 6 - 6 | 4101 | 67 | |
GenBank mRNA | AK055261.1 | - | 1 - 1 | 56 | 67 |
BC016029.1 | - | 6 - 6 | 3599 | 67 |