Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160541013-160541521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD84
Entrez Gene ID
8832
Gene Name
CD84 molecule
Gene Aliases
LY9B, SLAMF5, hCD84, mCD84
UniGene
Hs.398093
Gene Chromosome Location
Chr.1: 160541094-160579516 on Build GRCh38
Transcripts
NM_001184879.1 NM_001184881.1 NM_001184882.1 NM_003874.3 XM_011510094.2 XM_011510095.2

SNP Detail

SNP ID Chromosome Location
rs895905248 Chr.1: 160541042-160541042 on Build GRCh38
rs914932964 Chr.1: 160541291-160541291 on Build GRCh38
rs769640662 Chr.1: 160541172-160541172 on Build GRCh38
rs1018054887 Chr.1: 160541177-160541177 on Build GRCh38
rs1046025912 Chr.1: 160541180-160541180 on Build GRCh38
rs984178731 Chr.1: 160541058-160541058 on Build GRCh38
rs544207033 Chr.1: 160541198-160541198 on Build GRCh38
rs1005412260 Chr.1: 160541441-160541441 on Build GRCh38
rs575232970 Chr.1: 160541111-160541111 on Build GRCh38
rs994661905 Chr.1: 160541378-160541378 on Build GRCh38
rs1018125212 Chr.1: 160541492-160541492 on Build GRCh38
rs910004671 Chr.1: 160541065-160541070 on Build GRCh38
rs937466685 Chr.1: 160541342-160541342 on Build GRCh38
rs909955186 Chr.1: 160541387-160541387 on Build GRCh38
rs1043359174 Chr.1: 160541323-160541323 on Build GRCh38
rs897712063 Chr.1: 160541115-160541115 on Build GRCh38
rs1026071104 Chr.1: 160541074-160541074 on Build GRCh38
rs994151339 Chr.1: 160541066-160541066 on Build GRCh38
rs148915022 Chr.1: 160541040-160541040 on Build GRCh38
rs949297963 Chr.1: 160541443-160541443 on Build GRCh38
rs114612296 Chr.1: 160541395-160541395 on Build GRCh38
rs780983186 Chr.1: 160541060-160541060 on Build GRCh38
rs1001909028 Chr.1: 160541208-160541208 on Build GRCh38
rs1057331372 Chr.1: 160541364-160541364 on Build GRCh38
rs963874407 Chr.1: 160541201-160541201 on Build GRCh38
rs773100448 Chr.1: 160541251-160541251 on Build GRCh38
rs888479607 Chr.1: 160541404-160541404 on Build GRCh38
rs138063715 Chr.1: 160541374-160541374 on Build GRCh38
rs1053529507 Chr.1: 160541221-160541221 on Build GRCh38
rs145443123 Chr.1: 160541260-160541260 on Build GRCh38
rs770952347 Chr.1: 160541336-160541336 on Build GRCh38
rs574897598 Chr.1: 160541287-160541287 on Build GRCh38
rs542421177 Chr.1: 160541390-160541390 on Build GRCh38
rs114180564 Chr.1: 160541322-160541322 on Build GRCh38
rs527770316 Chr.1: 160541288-160541288 on Build GRCh38
rs568470248 Chr.1: 160541272-160541272 on Build GRCh38

View Full Product Details