Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160545935-160546468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD84
Entrez Gene ID
8832
Gene Name
CD84 molecule
Gene Aliases
LY9B, SLAMF5, hCD84, mCD84
UniGene
Hs.398093
Gene Chromosome Location
Chr.1: 160541094-160579516 on Build GRCh38
Transcripts
NM_001184879.1 NM_001184881.1 NM_001184882.1 NM_003874.3 XM_011510094.2 XM_011510095.2

SNP Detail

SNP ID Chromosome Location
rs1045172607 Chr.1: 160545967-160545967 on Build GRCh38
rs962521096 Chr.1: 160546354-160546354 on Build GRCh38
rs749465420 Chr.1: 160546306-160546306 on Build GRCh38
rs6686346 Chr.1: 160546023-160546023 on Build GRCh38
rs757600459 Chr.1: 160546416-160546416 on Build GRCh38
rs1006392075 Chr.1: 160546307-160546307 on Build GRCh38
rs1035509378 Chr.1: 160546402-160546402 on Build GRCh38
rs995769780 Chr.1: 160546293-160546293 on Build GRCh38
rs368271458 Chr.1: 160545991-160545992 on Build GRCh38
rs6662608 Chr.1: 160546004-160546004 on Build GRCh38
rs879038346 Chr.1: 160546208-160546208 on Build GRCh38
rs1048059369 Chr.1: 160546274-160546274 on Build GRCh38
rs568668192 Chr.1: 160546071-160546071 on Build GRCh38
rs1011543469 Chr.1: 160546344-160546344 on Build GRCh38
rs868081384 Chr.1: 160545975-160545975 on Build GRCh38
rs879570072 Chr.1: 160546335-160546335 on Build GRCh38
rs538040797 Chr.1: 160546091-160546091 on Build GRCh38
rs970383025 Chr.1: 160546223-160546223 on Build GRCh38
rs902039226 Chr.1: 160546287-160546287 on Build GRCh38
rs143943186 Chr.1: 160546141-160546141 on Build GRCh38
rs990168201 Chr.1: 160546191-160546191 on Build GRCh38
rs867963184 Chr.1: 160546005-160546005 on Build GRCh38
rs901514381 Chr.1: 160546068-160546068 on Build GRCh38
rs373495068 Chr.1: 160546235-160546235 on Build GRCh38
rs972478492 Chr.1: 160546257-160546257 on Build GRCh38
rs544052928 Chr.1: 160546294-160546294 on Build GRCh38
rs561180747 Chr.1: 160546345-160546345 on Build GRCh38
rs753884920 Chr.1: 160545990-160545991 on Build GRCh38
rs191624866 Chr.1: 160546230-160546230 on Build GRCh38
rs1018850926 Chr.1: 160546045-160546045 on Build GRCh38
rs867605965 Chr.1: 160546405-160546405 on Build GRCh38
rs56850015 Chr.1: 160546003-160546004 on Build GRCh38
rs948019876 Chr.1: 160546207-160546207 on Build GRCh38
rs938429414 Chr.1: 160546249-160546249 on Build GRCh38
rs969991443 Chr.1: 160546382-160546382 on Build GRCh38
rs539041458 Chr.1: 160546029-160546029 on Build GRCh38
rs929538993 Chr.1: 160546085-160546085 on Build GRCh38
rs928491447 Chr.1: 160546231-160546231 on Build GRCh38
rs1028686620 Chr.1: 160546084-160546084 on Build GRCh38
rs67091022 Chr.1: 160546004-160546004 on Build GRCh38
rs369638978 Chr.1: 160546268-160546268 on Build GRCh38
rs746856426 Chr.1: 160546276-160546276 on Build GRCh38
rs984953916 Chr.1: 160546133-160546133 on Build GRCh38
rs1018193408 Chr.1: 160546126-160546126 on Build GRCh38
rs930948813 Chr.1: 160546267-160546267 on Build GRCh38
rs546671570 Chr.1: 160546021-160546021 on Build GRCh38
rs969644844 Chr.1: 160546428-160546428 on Build GRCh38
rs1053170259 Chr.1: 160546317-160546317 on Build GRCh38
rs767640103 Chr.1: 160546092-160546092 on Build GRCh38
rs901795981 Chr.1: 160546290-160546290 on Build GRCh38
rs756206570 Chr.1: 160546247-160546247 on Build GRCh38
rs918213755 Chr.1: 160546182-160546182 on Build GRCh38
rs1031665949 Chr.1: 160546159-160546159 on Build GRCh38
rs998044576 Chr.1: 160546155-160546155 on Build GRCh38
rs745999368 Chr.1: 160546229-160546229 on Build GRCh38
rs6677895 Chr.1: 160546014-160546014 on Build GRCh38
rs569391449 Chr.1: 160546082-160546082 on Build GRCh38
rs943659459 Chr.1: 160546285-160546285 on Build GRCh38
rs559069185 Chr.1: 160546040-160546040 on Build GRCh38
rs903964814 Chr.1: 160545987-160545987 on Build GRCh38
rs1010576406 Chr.1: 160546042-160546042 on Build GRCh38
rs572458463 Chr.1: 160546251-160546251 on Build GRCh38
rs140199903 Chr.1: 160546108-160546108 on Build GRCh38
rs1001885154 Chr.1: 160546389-160546389 on Build GRCh38
rs992404241 Chr.1: 160546172-160546172 on Build GRCh38
rs887495778 Chr.1: 160546306-160546306 on Build GRCh38
rs1036646255 Chr.1: 160545972-160545972 on Build GRCh38
rs962287544 Chr.1: 160546162-160546162 on Build GRCh38
rs201124736 Chr.1: 160546022-160546023 on Build GRCh38
rs1025220583 Chr.1: 160546411-160546411 on Build GRCh38
rs115111574 Chr.1: 160546286-160546286 on Build GRCh38
rs1021674282 Chr.1: 160546360-160546360 on Build GRCh38
rs3766390 Chr.1: 160546259-160546259 on Build GRCh38
rs11548713 Chr.1: 160546160-160546160 on Build GRCh38
rs17381481 Chr.1: 160546355-160546355 on Build GRCh38

View Full Product Details