Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101031040-101031545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC17
Entrez Gene ID
140453
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
UniGene
Hs.271819
Gene Chromosome Location
Chr.7: 101020083-101058859 on Build GRCh38
Transcripts
NM_001040105.1 NR_133665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5966572 c.133delG p.D46fs*3 Chr.7: 101031170-101031170 on Build GRCh38
5793660 c.114A>T p.G38G Chr.7: 101031151-101031151 on Build GRCh38
6855663 c.88A>G p.S30G Chr.7: 101031125-101031125 on Build GRCh38
6277029 c.120G>T p.G40G Chr.7: 101031157-101031157 on Build GRCh38
5009100 c.127T>G p.S43A Chr.7: 101031164-101031164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139454683 Chr.7: 101031183-101031183 on Build GRCh38
rs557225784 Chr.7: 101031345-101031345 on Build GRCh38
rs746601415 Chr.7: 101031103-101031103 on Build GRCh38
rs906646298 Chr.7: 101031206-101031206 on Build GRCh38
rs761637007 Chr.7: 101031259-101031259 on Build GRCh38
rs915353374 Chr.7: 101031502-101031502 on Build GRCh38
rs749814718 Chr.7: 101031109-101031109 on Build GRCh38
rs879138631 Chr.7: 101031370-101031370 on Build GRCh38
rs949864054 Chr.7: 101031337-101031337 on Build GRCh38
rs1045026907 Chr.7: 101031227-101031227 on Build GRCh38
rs539996459 Chr.7: 101031456-101031456 on Build GRCh38
rs564010344 Chr.7: 101031347-101031347 on Build GRCh38
rs550305993 Chr.7: 101031176-101031176 on Build GRCh38
rs544640289 Chr.7: 101031299-101031299 on Build GRCh38
rs975484200 Chr.7: 101031079-101031079 on Build GRCh38
rs753700716 Chr.7: 101031165-101031165 on Build GRCh38
rs902784480 Chr.7: 101031489-101031489 on Build GRCh38
rs574919314 Chr.7: 101031504-101031504 on Build GRCh38
rs765209007 Chr.7: 101031175-101031175 on Build GRCh38
rs550586492 Chr.7: 101031435-101031435 on Build GRCh38
rs377521393 Chr.7: 101031252-101031252 on Build GRCh38
rs757826135 Chr.7: 101031095-101031095 on Build GRCh38
rs112089218 Chr.7: 101031262-101031262 on Build GRCh38
rs144369997 Chr.7: 101031186-101031186 on Build GRCh38
rs760752547 Chr.7: 101031148-101031148 on Build GRCh38
rs879156825 Chr.7: 101031325-101031325 on Build GRCh38
rs1055256217 Chr.7: 101031423-101031423 on Build GRCh38
rs985090027 Chr.7: 101031277-101031277 on Build GRCh38
rs776812072 Chr.7: 101031256-101031256 on Build GRCh38
rs1011574506 Chr.7: 101031222-101031222 on Build GRCh38
rs188591543 Chr.7: 101031328-101031328 on Build GRCh38
rs918535419 Chr.7: 101031321-101031321 on Build GRCh38
rs374703099 Chr.7: 101031211-101031211 on Build GRCh38
rs774981999 Chr.7: 101031111-101031111 on Build GRCh38
rs574004531 Chr.7: 101031283-101031283 on Build GRCh38
rs757121937 Chr.7: 101031089-101031089 on Build GRCh38
rs767459164 Chr.7: 101031118-101031118 on Build GRCh38
rs998777079 Chr.7: 101031368-101031368 on Build GRCh38
rs957262519 Chr.7: 101031449-101031449 on Build GRCh38
rs775368494 Chr.7: 101031233-101031233 on Build GRCh38
rs757034409 Chr.7: 101031171-101031171 on Build GRCh38
rs753790328 Chr.7: 101031085-101031085 on Build GRCh38
rs990387846 Chr.7: 101031485-101031485 on Build GRCh38
rs186080652 Chr.7: 101031453-101031453 on Build GRCh38
rs938102298 Chr.7: 101031377-101031377 on Build GRCh38
rs899575952 Chr.7: 101031293-101031293 on Build GRCh38
rs763883392 Chr.7: 101031155-101031155 on Build GRCh38
rs201971170 Chr.7: 101031185-101031185 on Build GRCh38
rs772208650 Chr.7: 101031232-101031232 on Build GRCh38
rs558418877 Chr.7: 101031240-101031240 on Build GRCh38
rs966853089 Chr.7: 101031235-101031235 on Build GRCh38
rs764360006 Chr.7: 101031253-101031253 on Build GRCh38
rs770506586 Chr.7: 101031239-101031241 on Build GRCh38
rs539367677 Chr.7: 101031268-101031268 on Build GRCh38
rs1021243270 Chr.7: 101031226-101031226 on Build GRCh38
rs1033765594 Chr.7: 101031284-101031284 on Build GRCh38
rs758208731 Chr.7: 101031269-101031269 on Build GRCh38
rs746315282 Chr.7: 101031230-101031230 on Build GRCh38
rs771630384 Chr.7: 101031110-101031110 on Build GRCh38
rs771397257 Chr.7: 101031221-101031221 on Build GRCh38
rs941075210 Chr.7: 101031282-101031282 on Build GRCh38
rs1038155958 Chr.7: 101031285-101031285 on Build GRCh38
rs779796548 Chr.7: 101031102-101031102 on Build GRCh38
rs909540476 Chr.7: 101031280-101031280 on Build GRCh38
rs770303716 Chr.7: 101031104-101031104 on Build GRCh38
rs998324048 Chr.7: 101031493-101031493 on Build GRCh38
rs778820128 Chr.7: 101031092-101031092 on Build GRCh38
rs759706435 Chr.7: 101031117-101031117 on Build GRCh38
rs775807508 Chr.7: 101031131-101031131 on Build GRCh38
rs1046446267 Chr.7: 101031190-101031190 on Build GRCh38
rs749724790 Chr.7: 101031212-101031212 on Build GRCh38
rs746853535 Chr.7: 101031081-101031082 on Build GRCh38
rs1021603633 Chr.7: 101031486-101031486 on Build GRCh38
rs140347706 Chr.7: 101031303-101031303 on Build GRCh38
rs749331352 Chr.7: 101031430-101031430 on Build GRCh38
rs745441771 Chr.7: 101031093-101031093 on Build GRCh38
rs894019743 Chr.7: 101031214-101031214 on Build GRCh38

View Full Product Details