Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLB1
Entrez Gene ID
151056
Gene Name
phospholipase B1
Gene Aliases
PLB, PLB/LIP, hPLB
Chromosome Location
Chr.2: 28488579 - 28644135 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170585.1 NP_001164056.1 20 - 21 1404 75
NM_153021.4 NP_694566.4 20 - 21 1371 75
XM_011532579.2 XP_011530881.1 22 - 23 1678 75
XM_011532581.2 XP_011530883.1 22 - 23 1641 75
XM_011532584.2 XP_011530886.1 22 - 23 1678 75
XM_011532588.2 XP_011530890.1 22 - 23 1680 75
XM_011532589.2 XP_011530891.1 22 - 23 1675 75
XM_011532590.2 XP_011530892.1 22 - 23 1680 75
XM_011532591.2 XP_011530893.1 22 - 23 1677 75
XM_011532593.2 XP_011530895.1 18 - 19 1253 75
XM_011532601.2 XP_011530903.1 8 - 9 666 75
XM_011532602.2 XP_011530904.1 7 - 8 638 75
XM_011532603.2 XP_011530905.1 7 - 8 715 75
XM_011532606.2 XP_011530908.1 22 - 23 1700 75
XM_011532607.2 XP_011530909.1 22 - 23 1700 75
XM_011532608.2 XP_011530910.1 22 - 23 1698 75
XM_011532609.2 XP_011530911.1 7 - 8 636 75
XM_017003432.1 XP_016858921.1 21 - 22 1647 75
XM_017003433.1 XP_016858922.1 21 - 22 1613 75
XM_017003434.1 XP_016858923.1 22 - 23 1682 75
GenBank mRNA AK055428.1 - 1 - 2 54 75
BC065041.1 - 7 - 8 457 75
BC150633.1 - 20 - 21 75
BC153864.1 - 20 - 21 1356 75

View Full Product Details