The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113511.2 | NP_001106983.1 | 20 - 20 | 3556 | 115 |
NM_001113512.2 | NP_001106984.1 | 18 - 18 | 3406 | 115 | |
NM_001320854.1 | NP_001307783.1 | 14 - 14 | 2018 | 115 | |
NM_145735.3 | NP_663788.1 | 19 - 19 | 3493 | 115 | |
XM_017020814.1 | XP_016876303.1 | 19 - 19 | 2657 | 115 | |
XM_017020817.1 | XP_016876306.1 | 17 - 17 | 2614 | 115 | |
XM_017020826.1 | XP_016876315.1 | 19 - 19 | 2489 | 115 | |
XM_017020827.1 | XP_016876316.1 | 17 - 17 | 2313 | 115 | |
XM_017020829.1 | XP_016876318.1 | 15 - 15 | 1937 | 115 | |
GenBank mRNA | AB075521.1 | - | 9 - 9 | 115 | |
AB177848.1 | - | 19 - 19 | 3493 | 115 | |
AK055476.1 | - | 1 - 1 | 50 | 115 | |
AK095058.1 | - | 4 - 4 | 2825 | 115 | |
AL834228.1 | - | 19 - 19 | 2677 | 115 | |
BC033905.1 | - | 18 - 18 | 2603 | 115 | |
BC060776.1 | - | 20 - 20 | 2898 | 115 | |
BX648314.1 | - | 1 - 1 | 4035 | 115 |