Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF12
Entrez Gene ID
7559
Gene Name
zinc finger protein 12
Gene Aliases
GIOT-3, HZF11, KOX3, ZNF325
Chromosome Location
Chr.7: 6688433 - 6706935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006956.2 NP_008887.2 6 - 6 4054 97
NM_016265.3 NP_057349.2 5 - 5 4168 97
GenBank mRNA AK055709.1 - 1 - 1 855 97
BC154414.1 - 5 - 5 4162 97
BX640749.1 - 5 - 5 3577 97

View Full Product Details