Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGCZ
Entrez Gene ID
137868
Gene Name
sarcoglycan zeta
Gene Aliases
ZSG1
Chromosome Location
Chr.8: 14084835 - 15238283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322879.1 NP_001309808.1 5 - 6 1178 82
NM_001322880.1 NP_001309809.1 5 - 6 1157 82
NM_001322881.1 NP_001309810.1 5 - 6 1063 82
NM_139167.3 NP_631906.2 6 - 7 1280 82
GenBank mRNA AY028700.1 - 5 - 6 581 82
AY502063.1 - 6 - 7 620 82
BC069459.1 - 5 - 6 581 82
BC125037.1 - 4 - 5 479 82

View Full Product Details