The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015089.3 | NP_055904.1 | 26 - 27 | 5286 | 71 |
XM_011514422.1 | XP_011512724.1 | 25 - 26 | 5442 | 71 | |
XM_011514423.2 | XP_011512725.1 | 26 - 27 | 5544 | 71 | |
XM_011514424.1 | XP_011512726.1 | 25 - 26 | 5361 | 71 | |
XM_011514425.1 | XP_011512727.1 | 23 - 24 | 5112 | 71 | |
XM_011514426.1 | XP_011512728.1 | 25 - 26 | 5442 | 71 | |
XM_011514427.1 | XP_011512729.1 | 21 - 22 | 4061 | 71 | |
XM_011514428.1 | XP_011512730.1 | 25 - 26 | 5442 | 71 | |
XM_011514429.1 | XP_011512731.1 | 18 - 19 | 3266 | 71 | |
XM_017010589.1 | XP_016866078.1 | 25 - 26 | 6153 | 71 | |
XM_017010590.1 | XP_016866079.1 | 19 - 20 | 3615 | 71 | |
GenBank mRNA | AB014608.3 | - | 18 - 19 | 2920 | 71 |
AJ318215.1 | - | 25 - 26 | 5211 | 71 | |
AY145132.1 | - | 25 - 26 | 5211 | 71 | |
BC002879.2 | - | 3 - 4 | 552 | 71 | |
CR749511.1 | - | 25 - 26 | 5665 | 71 | |
CR749841.1 | - | 26 - 27 | 5285 | 71 |