Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRDL1
Entrez Gene ID
91851
Gene Name
chordin like 1
Gene Aliases
CHL, MGC1, MGCN, NRLN1, VOPT, dA141H5.1
Chromosome Location
Chr.X: 110673856 - 110796058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143982.1 NP_001137454.1 4 - 5 652 79
NM_145234.3 NP_660277.2 4 - 5 500 79
XM_005262222.3 XP_005262279.1 4 - 5 469 79
XM_005262224.1 XP_005262281.1 4 - 5 468 79
GenBank mRNA AK312270.1 - 4 - 5 413 79
BC002909.2 - 4 - 5 417 79

View Full Product Details