Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG3
Entrez Gene ID
26030
Gene Name
pleckstrin homology and RhoGEF domain containing G3
Gene Aliases
ARHGEF43, KIAA0599
Chromosome Location
Chr.14: 64704129 - 64744346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308147.1 NP_001295076.1 16 - 17 3276 100
XM_011536624.2 XP_011534926.1 18 - 19 3557 100
XM_011536625.2 XP_011534927.1 18 - 19 3430 100
XM_011536627.2 XP_011534929.1 18 - 19 3822 100
XM_011536628.2 XP_011534930.1 18 - 19 4008 100
XM_011536629.2 XP_011534931.1 17 - 18 3146 100
XM_017021154.1 XP_016876643.1 17 - 18 3423 100
XM_017021155.1 XP_016876644.1 18 - 19 3512 100
XM_017021156.1 XP_016876645.1 18 - 19 3580 100
XM_017021157.1 XP_016876646.1 16 - 17 3376 100
XM_017021158.1 XP_016876647.1 16 - 17 3015 100
GenBank mRNA AB011171.1 - 1 - 2 1374 100
AK074169.1 - 10 - 11 3047 100
BC004298.1 - 4 - 5 1086 100
BC011891.2 - 1 - 2 1366 100
BC063554.1 - 2 - 3 1457 100
BC073907.1 - 3 - 4 1564 100
BC129952.1 - 10 - 11 2768 100
BC129953.1 - 16 - 17 3087 100

View Full Product Details