The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270707.1 | NP_001257636.1 | 3 - 4 | 674 | 116 |
NM_007243.2 | NP_009174.1 | 4 - 5 | 853 | 116 | |
NR_073065.1 | 3 - 4 | 678 | 116 | ||
NR_073066.1 | 2 - 3 | 459 | 116 | ||
XM_017010224.1 | XP_016865713.1 | 3 - 4 | 855 | 116 | |
XM_017010225.1 | XP_016865714.1 | 4 - 5 | 769 | 116 | |
GenBank mRNA | AF143676.1 | - | 3 - 4 | 499 | 116 |
AK075509.1 | - | 4 - 5 | 855 | 116 | |
AK298152.1 | - | 2 - 3 | 316 | 116 | |
AK315190.1 | - | 3 - 4 | 511 | 116 | |
AY358411.1 | - | 3 - 4 | 520 | 116 | |
BC004509.2 | - | 1 - 2 | 125 | 116 | |
BC021260.1 | - | 3 - 4 | 491 | 116 | |
BC039865.1 | - | 3 - 4 | 519 | 116 | |
BE548635.1 | - | 3 - 4 | 499 | 116 | |
BU159435.1 | - | 3 - 4 | 483 | 116 |