Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161047248-161047746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP30
Entrez Gene ID
257106
Gene Name
Rho GTPase activating protein 30
Gene Aliases
-
UniGene
Hs.389374
Gene Chromosome Location
Chr.1: 161046942-161069971 on Build GRCh38
Transcripts
NM_001025598.1 NM_001287600.1 NM_001287602.1 NM_181720.2 XM_005245070.2 XM_005245073.3 XM_011509391.2 XM_017000960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024271 c.2651C>A p.P884H Chr.1: 161047737-161047737 on Build GRCh38
4024270 c.3284C>A p.P1095H Chr.1: 161047737-161047737 on Build GRCh38
6603404 c.2659G>T p.G887C Chr.1: 161047729-161047729 on Build GRCh38
6603403 c.3292G>T p.G1098C Chr.1: 161047729-161047729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs544844350 Chr.1: 161047323-161047323 on Build GRCh38
rs186700346 Chr.1: 161047532-161047532 on Build GRCh38
rs765765530 Chr.1: 161047685-161047685 on Build GRCh38
rs181884600 Chr.1: 161047609-161047609 on Build GRCh38
rs773882907 Chr.1: 161047676-161047676 on Build GRCh38
rs567710980 Chr.1: 161047624-161047624 on Build GRCh38
rs984876262 Chr.1: 161047312-161047312 on Build GRCh38
rs75570999 Chr.1: 161047655-161047655 on Build GRCh38
rs146499279 Chr.1: 161047314-161047314 on Build GRCh38
rs374199320 Chr.1: 161047711-161047711 on Build GRCh38
rs1006730735 Chr.1: 161047617-161047617 on Build GRCh38
rs938450366 Chr.1: 161047579-161047579 on Build GRCh38
rs1057253002 Chr.1: 161047586-161047586 on Build GRCh38
rs989638796 Chr.1: 161047470-161047470 on Build GRCh38
rs372896794 Chr.1: 161047713-161047713 on Build GRCh38
rs757624964 Chr.1: 161047651-161047651 on Build GRCh38
rs377114513 Chr.1: 161047691-161047691 on Build GRCh38
rs544886366 Chr.1: 161047602-161047602 on Build GRCh38
rs567315346 Chr.1: 161047276-161047276 on Build GRCh38
rs536469739 Chr.1: 161047430-161047430 on Build GRCh38
rs35251010 Chr.1: 161047292-161047292 on Build GRCh38
rs1018677380 Chr.1: 161047338-161047338 on Build GRCh38
rs965501501 Chr.1: 161047407-161047407 on Build GRCh38
rs759224155 Chr.1: 161047679-161047679 on Build GRCh38
rs996425181 Chr.1: 161047580-161047580 on Build GRCh38
rs138342981 Chr.1: 161047650-161047650 on Build GRCh38
rs770420045 Chr.1: 161047672-161047672 on Build GRCh38
rs552945849 Chr.1: 161047519-161047519 on Build GRCh38
rs993999021 Chr.1: 161047595-161047595 on Build GRCh38
rs868717055 Chr.1: 161047523-161047523 on Build GRCh38
rs1044098036 Chr.1: 161047548-161047548 on Build GRCh38
rs907960905 Chr.1: 161047317-161047317 on Build GRCh38
rs979982057 Chr.1: 161047721-161047721 on Build GRCh38
rs935463832 Chr.1: 161047454-161047454 on Build GRCh38
rs956759388 Chr.1: 161047697-161047697 on Build GRCh38
rs1029111918 Chr.1: 161047600-161047600 on Build GRCh38
rs573666924 Chr.1: 161047437-161047437 on Build GRCh38
rs116809917 Chr.1: 161047612-161047612 on Build GRCh38
rs952129627 Chr.1: 161047302-161047302 on Build GRCh38
rs1018294556 Chr.1: 161047642-161047642 on Build GRCh38
rs896943347 Chr.1: 161047272-161047272 on Build GRCh38
rs745559856 Chr.1: 161047680-161047680 on Build GRCh38
rs34034855 Chr.1: 161047603-161047603 on Build GRCh38
rs1010287007 Chr.1: 161047531-161047531 on Build GRCh38
rs1031536635 Chr.1: 161047663-161047663 on Build GRCh38
rs750880413 Chr.1: 161047689-161047689 on Build GRCh38
rs145721081 Chr.1: 161047716-161047716 on Build GRCh38
rs1026894565 Chr.1: 161047279-161047279 on Build GRCh38
rs1054572516 Chr.1: 161047495-161047495 on Build GRCh38
rs902087241 Chr.1: 161047567-161047567 on Build GRCh38
rs371330926 Chr.1: 161047695-161047695 on Build GRCh38
rs566736875 Chr.1: 161047722-161047722 on Build GRCh38
rs367797654 Chr.1: 161047686-161047686 on Build GRCh38
rs1044247647 Chr.1: 161047451-161047451 on Build GRCh38
rs776040924 Chr.1: 161047700-161047701 on Build GRCh38
rs776959706 Chr.1: 161047683-161047683 on Build GRCh38
rs542565837 Chr.1: 161047460-161047460 on Build GRCh38
rs192492649 Chr.1: 161047591-161047591 on Build GRCh38
rs905502789 Chr.1: 161047554-161047554 on Build GRCh38
rs992191897 Chr.1: 161047335-161047335 on Build GRCh38

View Full Product Details