Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161509742-161510245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR2A
Entrez Gene ID
2212
Gene Name
Fc fragment of IgG receptor IIa
Gene Aliases
CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2
UniGene
Hs.352642
Gene Chromosome Location
Chr.1: 161505415-161524048 on Build GRCh38
Transcripts
NM_001136219.1 NM_021642.3 XM_011509287.2 XM_011509290.2 XM_011509291.1 XM_017000663.1 XM_017000664.1 XM_017000665.1 XM_017000666.1 XM_017000667.1 XM_017000668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6541155 c.365G>T p.W122L Chr.1: 161509823-161509823 on Build GRCh38
4924173 c.519C>T p.I173I Chr.1: 161509977-161509977 on Build GRCh38
3477276 c.403G>A p.G135R Chr.1: 161509861-161509861 on Build GRCh38
6662754 c.597T>C p.P199P Chr.1: 161510055-161510055 on Build GRCh38
6122134 c.578C>A p.T193K Chr.1: 161510036-161510036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770453527 Chr.1: 161509967-161509967 on Build GRCh38
rs773620534 Chr.1: 161509909-161509909 on Build GRCh38
rs145979252 Chr.1: 161509918-161509918 on Build GRCh38
rs989024887 Chr.1: 161509885-161509885 on Build GRCh38
rs781127278 Chr.1: 161509946-161509946 on Build GRCh38
rs764458028 Chr.1: 161509890-161509890 on Build GRCh38
rs868229314 Chr.1: 161509963-161509963 on Build GRCh38
rs377303335 Chr.1: 161510019-161510019 on Build GRCh38
rs776005198 Chr.1: 161509884-161509884 on Build GRCh38
rs748029135 Chr.1: 161509950-161509950 on Build GRCh38
rs776769021 Chr.1: 161509891-161509891 on Build GRCh38
rs944977866 Chr.1: 161509892-161509892 on Build GRCh38
rs141094947 Chr.1: 161510042-161510042 on Build GRCh38
rs550128935 Chr.1: 161510142-161510142 on Build GRCh38
rs779490441 Chr.1: 161509903-161509903 on Build GRCh38
rs940504688 Chr.1: 161510015-161510015 on Build GRCh38
rs201829744 Chr.1: 161510120-161510120 on Build GRCh38
rs769218774 Chr.1: 161510027-161510027 on Build GRCh38
rs748874961 Chr.1: 161509869-161509869 on Build GRCh38
rs139202723 Chr.1: 161509942-161509942 on Build GRCh38
rs754079714 Chr.1: 161509786-161509786 on Build GRCh38
rs557989931 Chr.1: 161510173-161510173 on Build GRCh38
rs947911066 Chr.1: 161510203-161510203 on Build GRCh38
rs567744828 Chr.1: 161510099-161510099 on Build GRCh38
rs150311303 Chr.1: 161510070-161510071 on Build GRCh38
rs4986941 Chr.1: 161509873-161509873 on Build GRCh38
rs764257889 Chr.1: 161509782-161509782 on Build GRCh38
rs754843836 Chr.1: 161509834-161509834 on Build GRCh38
rs141572415 Chr.1: 161509848-161509848 on Build GRCh38
rs111242375 Chr.1: 161509914-161509914 on Build GRCh38
rs750582877 Chr.1: 161509802-161509802 on Build GRCh38
rs1033051928 Chr.1: 161510218-161510218 on Build GRCh38
rs540989857 Chr.1: 161510206-161510206 on Build GRCh38
rs7529425 Chr.1: 161509809-161509809 on Build GRCh38
rs777342855 Chr.1: 161509930-161509930 on Build GRCh38
rs558084508 Chr.1: 161510212-161510212 on Build GRCh38
rs912139222 Chr.1: 161509889-161509889 on Build GRCh38
rs774829021 Chr.1: 161509958-161509958 on Build GRCh38
rs138950286 Chr.1: 161510064-161510064 on Build GRCh38
rs368752463 Chr.1: 161510031-161510031 on Build GRCh38
rs1056150031 Chr.1: 161510186-161510186 on Build GRCh38
rs149603892 Chr.1: 161509886-161509886 on Build GRCh38
rs79273228 Chr.1: 161510037-161510037 on Build GRCh38
rs976279797 Chr.1: 161509800-161509800 on Build GRCh38
rs375004817 Chr.1: 161510122-161510122 on Build GRCh38
rs1801274 Chr.1: 161509955-161509955 on Build GRCh38
rs751753520 Chr.1: 161510049-161510049 on Build GRCh38
rs375025584 Chr.1: 161509851-161509851 on Build GRCh38
rs55722329 Chr.1: 161510070-161510070 on Build GRCh38
rs760172471 Chr.1: 161509991-161509991 on Build GRCh38
rs371752364 Chr.1: 161509825-161509825 on Build GRCh38
rs956387905 Chr.1: 161509881-161509881 on Build GRCh38
rs372733941 Chr.1: 161509980-161509980 on Build GRCh38
rs747763665 Chr.1: 161509841-161509841 on Build GRCh38
rs757302933 Chr.1: 161509788-161509788 on Build GRCh38
rs896295020 Chr.1: 161510199-161510199 on Build GRCh38
rs930285323 Chr.1: 161510127-161510127 on Build GRCh38
rs754949042 Chr.1: 161510052-161510052 on Build GRCh38
rs555143591 Chr.1: 161510165-161510165 on Build GRCh38
rs748710561 Chr.1: 161509964-161509964 on Build GRCh38
rs778861856 Chr.1: 161509791-161509791 on Build GRCh38
rs199737051 Chr.1: 161509829-161509829 on Build GRCh38
rs745440888 Chr.1: 161510096-161510096 on Build GRCh38
rs769130420 Chr.1: 161510097-161510097 on Build GRCh38
rs535567676 Chr.1: 161510102-161510102 on Build GRCh38
rs572025603 Chr.1: 161510169-161510169 on Build GRCh38
rs765553715 Chr.1: 161509902-161509902 on Build GRCh38
rs752407793 Chr.1: 161510068-161510068 on Build GRCh38
rs763066799 Chr.1: 161510040-161510040 on Build GRCh38
rs55919520 Chr.1: 161510080-161510080 on Build GRCh38
rs778391223 Chr.1: 161510083-161510083 on Build GRCh38
rs141159775 Chr.1: 161509920-161509920 on Build GRCh38
rs765336574 Chr.1: 161510036-161510036 on Build GRCh38
rs571568091 Chr.1: 161510069-161510069 on Build GRCh38
rs780937135 Chr.1: 161509838-161509838 on Build GRCh38
rs761938152 Chr.1: 161509899-161509899 on Build GRCh38
rs373950032 Chr.1: 161509780-161509780 on Build GRCh38

View Full Product Details