Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPGS1
Entrez Gene ID
91978
Gene Name
tubulin polyglutamylase complex subunit 1
Gene Aliases
C19orf20, GTRGEO22, PGs1
Chromosome Location
Chr.19: 507497 - 519654 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033513.2 NP_277048.2 1 - 2 347 68
GenBank mRNA AK126170.1 - 1 - 2 347 68
BC009520.1 - 1 - 2 233 68
BM846833.1 - 1 - 2 347 68

View Full Product Details