The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001872.3 | NP_001001872.2 | 3 - 4 | 1846 | 72 |
NM_001320173.1 | NP_001307102.1 | 3 - 4 | 1846 | 72 | |
XM_011536471.2 | XP_011534773.1 | 3 - 4 | 1846 | 72 | |
XM_017021029.1 | XP_016876518.1 | 3 - 4 | 1703 | 72 | |
XM_017021030.1 | XP_016876519.1 | 3 - 4 | 1846 | 72 | |
GenBank mRNA | AK292513.1 | - | 3 - 4 | 1820 | 72 |
AK292605.1 | - | 3 - 4 | 1763 | 72 | |
AK294599.1 | - | 2 - 3 | 1057 | 72 | |
BC013707.1 | - | 2 - 3 | 1143 | 72 | |
BC146861.1 | - | 3 - 4 | 1697 | 72 | |
BX161411.1 | - | 3 - 4 | 1736 | 72 | |
BX161448.1 | - | 3 - 4 | 1790 | 72 | |
BX649130.1 | - | 2 - 3 | 1342 | 72 |