Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86152815-86153334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0D2
Entrez Gene ID
245972
Gene Name
ATPase H+ transporting V0 subunit d2
Gene Aliases
ATP6D2, VMA6
UniGene
Hs.436360
Gene Chromosome Location
Chr.8: 86098910-86154225 on Build GRCh38
Transcripts
NM_152565.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6114212 c.898A>T p.M300L Chr.8: 86152822-86152822 on Build GRCh38
197703 c.1019G>A p.R340Q Chr.8: 86152943-86152943 on Build GRCh38
2789563 c.897A>T p.Q299H Chr.8: 86152821-86152821 on Build GRCh38
454905 c.900G>C p.M300I Chr.8: 86152824-86152824 on Build GRCh38
5455691 c.934G>A p.G312S Chr.8: 86152858-86152858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889059664 Chr.8: 86153102-86153102 on Build GRCh38
rs775719901 Chr.8: 86152858-86152858 on Build GRCh38
rs1011237392 Chr.8: 86153185-86153185 on Build GRCh38
rs1052792615 Chr.8: 86153114-86153114 on Build GRCh38
rs747659877 Chr.8: 86152840-86152840 on Build GRCh38
rs780447559 Chr.8: 86152993-86152993 on Build GRCh38
rs772542257 Chr.8: 86153000-86153000 on Build GRCh38
rs916851570 Chr.8: 86152916-86152916 on Build GRCh38
rs137869896 Chr.8: 86152929-86152929 on Build GRCh38
rs892899673 Chr.8: 86153154-86153154 on Build GRCh38
rs923128345 Chr.8: 86153072-86153072 on Build GRCh38
rs993251090 Chr.8: 86153190-86153190 on Build GRCh38
rs1050831017 Chr.8: 86153089-86153089 on Build GRCh38
rs546966931 Chr.8: 86153121-86153121 on Build GRCh38
rs751331505 Chr.8: 86152919-86152919 on Build GRCh38
rs777431472 Chr.8: 86152963-86152963 on Build GRCh38
rs910462020 Chr.8: 86153020-86153020 on Build GRCh38
rs761202061 Chr.8: 86152864-86152864 on Build GRCh38
rs943298653 Chr.8: 86153051-86153051 on Build GRCh38
rs1037571460 Chr.8: 86153181-86153181 on Build GRCh38
rs765436243 Chr.8: 86152893-86152893 on Build GRCh38
rs935478518 Chr.8: 86152979-86152979 on Build GRCh38
rs566879130 Chr.8: 86153209-86153209 on Build GRCh38
rs530035423 Chr.8: 86153062-86153062 on Build GRCh38
rs968361022 Chr.8: 86153283-86153283 on Build GRCh38
rs748880537 Chr.8: 86152964-86152964 on Build GRCh38
rs769094980 Chr.8: 86152865-86152865 on Build GRCh38
rs142355302 Chr.8: 86152842-86152842 on Build GRCh38
rs1052561976 Chr.8: 86153048-86153052 on Build GRCh38
rs370435789 Chr.8: 86153302-86153302 on Build GRCh38
rs960028619 Chr.8: 86153239-86153239 on Build GRCh38
rs757035613 Chr.8: 86152845-86152845 on Build GRCh38
rs756479018 Chr.8: 86153019-86153019 on Build GRCh38
rs906078768 Chr.8: 86153222-86153222 on Build GRCh38
rs376070860 Chr.8: 86152857-86152857 on Build GRCh38
rs1003129366 Chr.8: 86153275-86153275 on Build GRCh38
rs770735466 Chr.8: 86152988-86152988 on Build GRCh38
rs568342882 Chr.8: 86153230-86153231 on Build GRCh38
rs769204928 Chr.8: 86153007-86153007 on Build GRCh38
rs35552884 Chr.8: 86153184-86153185 on Build GRCh38
rs148275781 Chr.8: 86153235-86153235 on Build GRCh38
rs146069775 Chr.8: 86152913-86152913 on Build GRCh38
rs761951622 Chr.8: 86152872-86152872 on Build GRCh38
rs781030035 Chr.8: 86152943-86152943 on Build GRCh38
rs755739722 Chr.8: 86152947-86152947 on Build GRCh38
rs754919411 Chr.8: 86152942-86152942 on Build GRCh38
rs1035452198 Chr.8: 86153225-86153225 on Build GRCh38
rs746547441 Chr.8: 86153013-86153013 on Build GRCh38
rs752246824 Chr.8: 86152946-86152946 on Build GRCh38
rs561469026 Chr.8: 86153023-86153023 on Build GRCh38
rs531159801 Chr.8: 86152870-86152870 on Build GRCh38
rs1044485581 Chr.8: 86153217-86153217 on Build GRCh38
rs934384577 Chr.8: 86153091-86153091 on Build GRCh38
rs777066606 Chr.8: 86153008-86153008 on Build GRCh38
rs773066311 Chr.8: 86152854-86152854 on Build GRCh38
rs970752253 Chr.8: 86152851-86152851 on Build GRCh38
rs750586607 Chr.8: 86152900-86152900 on Build GRCh38
rs763168244 Chr.8: 86152905-86152905 on Build GRCh38
rs780274242 Chr.8: 86153054-86153054 on Build GRCh38
rs1012602354 Chr.8: 86153273-86153273 on Build GRCh38
rs867046104 Chr.8: 86153022-86153022 on Build GRCh38
rs552520868 Chr.8: 86153304-86153304 on Build GRCh38
rs762322021 Chr.8: 86153013-86153013 on Build GRCh38
rs747376053 Chr.8: 86152997-86152997 on Build GRCh38

View Full Product Details