Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86575416-86575945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
UniGene
Hs.154433
Gene Chromosome Location
Chr.8: 86573608-86743675 on Build GRCh38
Transcripts
NM_019098.4 XM_011517138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703443 c.2344C>G p.Q782E Chr.8: 86575890-86575890 on Build GRCh38
3651514 c.2386G>A p.E796K Chr.8: 86575848-86575848 on Build GRCh38
319528 c.2368C>G p.P790A Chr.8: 86575866-86575866 on Build GRCh38
6549505 c.2328C>A p.P776P Chr.8: 86575906-86575906 on Build GRCh38
6114194 c.2301_2302CC>AT p.H768Y Chr.8: 86575932-86575933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142846289 Chr.8: 86575814-86575814 on Build GRCh38
rs753083465 Chr.8: 86575851-86575851 on Build GRCh38
rs774763662 Chr.8: 86575799-86575799 on Build GRCh38
rs566764255 Chr.8: 86575737-86575737 on Build GRCh38
rs753199958 Chr.8: 86575839-86575839 on Build GRCh38
rs758053992 Chr.8: 86575845-86575845 on Build GRCh38
rs760477374 Chr.8: 86575816-86575816 on Build GRCh38
rs749307247 Chr.8: 86575867-86575867 on Build GRCh38
rs189210452 Chr.8: 86575753-86575753 on Build GRCh38
rs758914061 Chr.8: 86575919-86575921 on Build GRCh38
rs1031324821 Chr.8: 86575696-86575696 on Build GRCh38
rs141717983 Chr.8: 86575511-86575511 on Build GRCh38
rs750147398 Chr.8: 86575842-86575842 on Build GRCh38
rs372823505 Chr.8: 86575872-86575872 on Build GRCh38
rs138432513 Chr.8: 86575775-86575775 on Build GRCh38
rs775381466 Chr.8: 86575893-86575893 on Build GRCh38
rs981845761 Chr.8: 86575533-86575533 on Build GRCh38
rs957300863 Chr.8: 86575706-86575706 on Build GRCh38
rs369349856 Chr.8: 86575804-86575804 on Build GRCh38
rs561845227 Chr.8: 86575802-86575802 on Build GRCh38
rs945309818 Chr.8: 86575852-86575852 on Build GRCh38
rs886063159 Chr.8: 86575598-86575598 on Build GRCh38
rs372938106 Chr.8: 86575720-86575720 on Build GRCh38
rs770350464 Chr.8: 86575778-86575778 on Build GRCh38
rs1040581292 Chr.8: 86575607-86575607 on Build GRCh38
rs116597007 Chr.8: 86575636-86575636 on Build GRCh38
rs761282321 Chr.8: 86575785-86575785 on Build GRCh38
rs150149878 Chr.8: 86575537-86575537 on Build GRCh38
rs772725745 Chr.8: 86575912-86575912 on Build GRCh38
rs770198026 Chr.8: 86575523-86575523 on Build GRCh38
rs772444831 Chr.8: 86575892-86575892 on Build GRCh38
rs1027817115 Chr.8: 86575724-86575724 on Build GRCh38
rs375288585 Chr.8: 86575815-86575815 on Build GRCh38
rs764860583 Chr.8: 86575837-86575837 on Build GRCh38
rs186448979 Chr.8: 86575819-86575819 on Build GRCh38
rs764662692 Chr.8: 86575909-86575909 on Build GRCh38
rs937844558 Chr.8: 86575553-86575553 on Build GRCh38
rs989289169 Chr.8: 86575564-86575565 on Build GRCh38
rs776684960 Chr.8: 86575900-86575900 on Build GRCh38
rs750535143 Chr.8: 86575884-86575884 on Build GRCh38
rs773875783 Chr.8: 86575777-86575777 on Build GRCh38
rs1039114842 Chr.8: 86575714-86575714 on Build GRCh38
rs760554096 Chr.8: 86575895-86575895 on Build GRCh38
rs762547089 Chr.8: 86575918-86575918 on Build GRCh38
rs999117032 Chr.8: 86575881-86575881 on Build GRCh38
rs761227251 Chr.8: 86575908-86575908 on Build GRCh38
rs1042820229 Chr.8: 86575637-86575637 on Build GRCh38
rs762595113 Chr.8: 86575728-86575728 on Build GRCh38
rs545964518 Chr.8: 86575454-86575454 on Build GRCh38
rs777666964 Chr.8: 86575784-86575784 on Build GRCh38
rs930742286 Chr.8: 86575455-86575455 on Build GRCh38
rs867410377 Chr.8: 86575710-86575710 on Build GRCh38
rs537918624 Chr.8: 86575788-86575788 on Build GRCh38
rs944713089 Chr.8: 86575886-86575886 on Build GRCh38
rs267602028 Chr.8: 86575869-86575869 on Build GRCh38
rs1049105014 Chr.8: 86575823-86575823 on Build GRCh38
rs781664615 Chr.8: 86575764-86575764 on Build GRCh38
rs140932384 Chr.8: 86575859-86575859 on Build GRCh38
rs778384043 Chr.8: 86575866-86575866 on Build GRCh38
rs779201314 Chr.8: 86575777-86575778 on Build GRCh38
rs768453158 Chr.8: 86575898-86575898 on Build GRCh38
rs1007696057 Chr.8: 86575575-86575575 on Build GRCh38
rs781481819 Chr.8: 86575847-86575847 on Build GRCh38
rs961305731 Chr.8: 86575809-86575809 on Build GRCh38
rs900557831 Chr.8: 86575719-86575719 on Build GRCh38
rs186370374 Chr.8: 86575679-86575679 on Build GRCh38
rs543097492 Chr.8: 86575657-86575657 on Build GRCh38
rs181350408 Chr.8: 86575516-86575516 on Build GRCh38
rs886731109 Chr.8: 86575727-86575727 on Build GRCh38
rs151039691 Chr.8: 86575824-86575824 on Build GRCh38
rs201365222 Chr.8: 86575787-86575787 on Build GRCh38
rs753251909 Chr.8: 86575821-86575821 on Build GRCh38

View Full Product Details