The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012287.5 | NP_036419.3 | 1 - 2 | 295 | 70 |
XM_006713557.3 | XP_006713620.1 | 1 - 2 | 295 | 70 | |
XM_011512602.2 | XP_011510904.1 | 1 - 2 | 295 | 70 | |
XM_011512603.2 | XP_011510905.1 | 1 - 2 | 295 | 70 | |
XM_017006046.1 | XP_016861535.1 | 1 - 2 | 319 | 70 | |
XM_017006047.1 | XP_016861536.1 | 1 - 2 | 319 | 70 | |
XM_017006048.1 | XP_016861537.1 | 1 - 2 | 319 | 70 | |
XM_017006049.1 | XP_016861538.1 | 1 - 2 | 319 | 70 | |
GenBank mRNA | AJ238248.1 | - | 1 - 2 | 114 | 70 |
AK222635.1 | - | 1 - 2 | 250 | 70 | |
AK290369.1 | - | 1 - 2 | 227 | 70 | |
BC060767.1 | - | 1 - 2 | 230 | 70 | |
D26069.3 | - | 1 - 2 | 64 | 70 | |
DA774230.1 | - | 1 - 2 | 250 | 70 | |
DB087306.1 | - | 1 - 2 | 295 | 70 |