The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017580.2 | NP_060050.2 | 2 - 3 | 1372 | 69 |
XM_005269925.4 | XP_005269982.1 | 3 - 4 | 1227 | 69 | |
XM_005269926.4 | XP_005269983.1 | 3 - 4 | 1415 | 69 | |
XM_006717907.2 | XP_006717970.1 | 3 - 4 | 2097 | 69 | |
XM_017016357.1 | XP_016871846.1 | 3 - 4 | 2828 | 69 | |
XM_017016358.1 | XP_016871847.1 | 3 - 4 | 1062 | 69 | |
GenBank mRNA | AJ252060.1 | - | 2 - 3 | 1407 | 69 |
AK302810.1 | - | 3 - 4 | 1183 | 69 | |
AK307386.1 | - | 2 - 3 | 1012 | 69 | |
AL832925.1 | - | 2 - 3 | 552 | 69 |