Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168079837-168080337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR161
Entrez Gene ID
23432
Gene Name
G protein-coupled receptor 161
Gene Aliases
RE2
UniGene
Hs.271809
Gene Chromosome Location
Chr.1: 168079542-168137667 on Build GRCh38
Transcripts
NM_001267609.1 NM_001267610.1 NM_001267611.1 NM_001267612.1 NM_001267613.1 NM_001267614.1 NM_153832.2 XM_005245055.2 XM_005245056.2 XM_005245057.4 XM_006711251.1 XM_006711253.2 XM_011509370.1 XM_011509371.2 XM_011509372.1 XM_011509373.2 XM_011509375.2 XM_011509376.2 XM_011509377.1 XM_011509378.1 XM_017000858.1 XM_017000859.1 XM_017000860.1 XM_017000861.1 XM_017000862.1

SNP Detail

SNP ID Chromosome Location
rs575595836 Chr.1: 168080288-168080288 on Build GRCh38
rs1018046512 Chr.1: 168080238-168080238 on Build GRCh38
rs917799071 Chr.1: 168080046-168080046 on Build GRCh38
rs181631923 Chr.1: 168079993-168079993 on Build GRCh38
rs35017771 Chr.1: 168080023-168080024 on Build GRCh38
rs762034247 Chr.1: 168080205-168080205 on Build GRCh38
rs866092678 Chr.1: 168080110-168080110 on Build GRCh38
rs3795606 Chr.1: 168080095-168080095 on Build GRCh38
rs529051306 Chr.1: 168080272-168080272 on Build GRCh38
rs937775679 Chr.1: 168080008-168080008 on Build GRCh38
rs951224901 Chr.1: 168079957-168079957 on Build GRCh38
rs563727954 Chr.1: 168080191-168080191 on Build GRCh38
rs374258820 Chr.1: 168080003-168080003 on Build GRCh38
rs765701098 Chr.1: 168080268-168080268 on Build GRCh38
rs933492080 Chr.1: 168080136-168080136 on Build GRCh38
rs1019437457 Chr.1: 168080174-168080174 on Build GRCh38
rs867594811 Chr.1: 168080096-168080096 on Build GRCh38
rs572832623 Chr.1: 168080196-168080196 on Build GRCh38
rs1052399100 Chr.1: 168080167-168080167 on Build GRCh38
rs991445086 Chr.1: 168080062-168080062 on Build GRCh38
rs10918822 Chr.1: 168079991-168079991 on Build GRCh38
rs75717090 Chr.1: 168080178-168080178 on Build GRCh38
rs79611045 Chr.1: 168080181-168080181 on Build GRCh38
rs545438116 Chr.1: 168080187-168080187 on Build GRCh38
rs991957150 Chr.1: 168080256-168080256 on Build GRCh38
rs760998945 Chr.1: 168080197-168080197 on Build GRCh38
rs952288754 Chr.1: 168080200-168080200 on Build GRCh38
rs945197256 Chr.1: 168080048-168080048 on Build GRCh38
rs141180854 Chr.1: 168080294-168080294 on Build GRCh38
rs772195623 Chr.1: 168079942-168079942 on Build GRCh38
rs1042189392 Chr.1: 168080051-168080051 on Build GRCh38
rs947398510 Chr.1: 168080075-168080075 on Build GRCh38
rs565143178 Chr.1: 168080127-168080130 on Build GRCh38
rs914534959 Chr.1: 168080072-168080072 on Build GRCh38
rs368097572 Chr.1: 168080121-168080121 on Build GRCh38
rs769059125 Chr.1: 168080202-168080209 on Build GRCh38
rs1056599755 Chr.1: 168080041-168080041 on Build GRCh38
rs903783004 Chr.1: 168080065-168080065 on Build GRCh38
rs1007681789 Chr.1: 168080170-168080171 on Build GRCh38
rs1019133667 Chr.1: 168079867-168079867 on Build GRCh38
rs966607427 Chr.1: 168079869-168079869 on Build GRCh38
rs79718642 Chr.1: 168080179-168080179 on Build GRCh38
rs995192365 Chr.1: 168080253-168080253 on Build GRCh38
rs926301994 Chr.1: 168080240-168080240 on Build GRCh38
rs972375481 Chr.1: 168079874-168079874 on Build GRCh38
rs561754658 Chr.1: 168080264-168080264 on Build GRCh38
rs985031085 Chr.1: 168080223-168080223 on Build GRCh38
rs926251114 Chr.1: 168080006-168080006 on Build GRCh38
rs984472316 Chr.1: 168080005-168080005 on Build GRCh38
rs1002700616 Chr.1: 168080306-168080306 on Build GRCh38
rs147800244 Chr.1: 168079866-168079866 on Build GRCh38
rs945107021 Chr.1: 168080279-168080280 on Build GRCh38
rs113119901 Chr.1: 168080250-168080250 on Build GRCh38
rs917667334 Chr.1: 168080257-168080257 on Build GRCh38

View Full Product Details