Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 168084981-168085484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR161
Entrez Gene ID
23432
Gene Name
G protein-coupled receptor 161
Gene Aliases
RE2
UniGene
Hs.271809
Gene Chromosome Location
Chr.1: 168079542-168137667 on Build GRCh38
Transcripts
NM_001267609.1 NM_001267610.1 NM_001267611.1 NM_001267612.1 NM_001267613.1 NM_001267614.1 NM_153832.2 XM_005245055.2 XM_005245056.2 XM_005245057.4 XM_006711251.1 XM_006711253.2 XM_011509370.1 XM_011509371.2 XM_011509372.1 XM_011509373.2 XM_011509375.2 XM_011509376.2 XM_011509377.1 XM_011509378.1 XM_017000858.1 XM_017000859.1 XM_017000860.1 XM_017000861.1 XM_017000862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4986037 c.59T>C p.L20P Chr.1: 168085161-168085161 on Build GRCh38
3849315 c.36C>T p.D12D Chr.1: 168085184-168085184 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs934027122 Chr.1: 168085241-168085241 on Build GRCh38
rs570659920 Chr.1: 168085046-168085046 on Build GRCh38
rs275139 Chr.1: 168085162-168085162 on Build GRCh38
rs375020953 Chr.1: 168085231-168085232 on Build GRCh38
rs568423127 Chr.1: 168085159-168085159 on Build GRCh38
rs915165591 Chr.1: 168085175-168085175 on Build GRCh38
rs768622853 Chr.1: 168085003-168085004 on Build GRCh38
rs1034850157 Chr.1: 168085202-168085202 on Build GRCh38
rs1049798837 Chr.1: 168085019-168085019 on Build GRCh38
rs12567718 Chr.1: 168085265-168085265 on Build GRCh38
rs931790003 Chr.1: 168085033-168085033 on Build GRCh38
rs979422187 Chr.1: 168085256-168085257 on Build GRCh38
rs748509904 Chr.1: 168085180-168085180 on Build GRCh38
rs535110879 Chr.1: 168085093-168085093 on Build GRCh38
rs142320164 Chr.1: 168085237-168085238 on Build GRCh38
rs977955118 Chr.1: 168085197-168085197 on Build GRCh38
rs770237897 Chr.1: 168085427-168085427 on Build GRCh38
rs1038600045 Chr.1: 168085324-168085324 on Build GRCh38
rs12568392 Chr.1: 168085267-168085267 on Build GRCh38
rs1014744388 Chr.1: 168085065-168085065 on Build GRCh38
rs768737647 Chr.1: 168085050-168085050 on Build GRCh38
rs373389874 Chr.1: 168085183-168085183 on Build GRCh38
rs371644972 Chr.1: 168085092-168085092 on Build GRCh38
rs889795919 Chr.1: 168085026-168085026 on Build GRCh38
rs762253160 Chr.1: 168085268-168085268 on Build GRCh38
rs191098984 Chr.1: 168085203-168085203 on Build GRCh38
rs558374051 Chr.1: 168085327-168085327 on Build GRCh38
rs376651598 Chr.1: 168085423-168085423 on Build GRCh38
rs1008740500 Chr.1: 168085044-168085044 on Build GRCh38
rs541098922 Chr.1: 168085397-168085397 on Build GRCh38
rs901289021 Chr.1: 168085005-168085005 on Build GRCh38
rs117550517 Chr.1: 168085240-168085240 on Build GRCh38
rs574341444 Chr.1: 168085457-168085457 on Build GRCh38
rs931649532 Chr.1: 168085455-168085455 on Build GRCh38
rs1033272686 Chr.1: 168085154-168085154 on Build GRCh38
rs904169451 Chr.1: 168085027-168085027 on Build GRCh38
rs941378835 Chr.1: 168085321-168085321 on Build GRCh38
rs548630394 Chr.1: 168085106-168085108 on Build GRCh38
rs73028189 Chr.1: 168085095-168085095 on Build GRCh38
rs559107961 Chr.1: 168085431-168085431 on Build GRCh38
rs557644479 Chr.1: 168085174-168085174 on Build GRCh38
rs41271649 Chr.1: 168085038-168085038 on Build GRCh38
rs992969010 Chr.1: 168085221-168085222 on Build GRCh38
rs907389257 Chr.1: 168085437-168085437 on Build GRCh38
rs375166676 Chr.1: 168085136-168085136 on Build GRCh38
rs897299054 Chr.1: 168085086-168085086 on Build GRCh38
rs765851231 Chr.1: 168085229-168085229 on Build GRCh38
rs112973198 Chr.1: 168085267-168085268 on Build GRCh38
rs1051426 Chr.1: 168085035-168085035 on Build GRCh38
rs367867487 Chr.1: 168085012-168085012 on Build GRCh38
rs575911565 Chr.1: 168085177-168085177 on Build GRCh38
rs1024388755 Chr.1: 168085416-168085416 on Build GRCh38
rs73028190 Chr.1: 168085393-168085393 on Build GRCh38
rs967627930 Chr.1: 168085246-168085246 on Build GRCh38
rs189511250 Chr.1: 168085129-168085129 on Build GRCh38

View Full Product Details