The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166271.2 | NP_001159743.1 | 13 - 13 | 7804 | 149 |
NM_001286792.1 | NP_001273721.1 | 15 - 15 | 8128 | 149 | |
NM_001286793.1 | NP_001273722.1 | 9 - 9 | 5352 | 149 | |
NM_001286794.1 | NP_001273723.1 | 11 - 11 | 5746 | 149 | |
NM_001286795.1 | NP_001273724.1 | 10 - 10 | 5538 | 149 | |
NM_153023.3 | NP_694568.1 | 12 - 12 | 6040 | 149 | |
GenBank mRNA | AK025586.1 | - | 1 - 1 | 1390 | 149 |
AK074117.1 | - | 3 - 3 | 4250 | 149 | |
AK160371.1 | - | 9 - 9 | 5227 | 149 | |
AL049335.1 | - | 1 - 1 | 342 | 149 | |
BX648244.1 | - | 15 - 15 | 7950 | 149 |