The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142523.1 | NP_001135995.1 | 11 - 11 | 5917 | 63 |
NM_007199.2 | NP_009130.2 | 12 - 12 | 6100 | 63 | |
GenBank mRNA | AK023663.1 | - | 1 - 1 | 1040 | 63 |
AL049435.1 | - | 1 - 1 | 451 | 63 |