Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCHY1
Entrez Gene ID
25898
Gene Name
ring finger and CHY zinc finger domain containing 1
Gene Aliases
ARNIP, CHIMP, PIRH2, PRO1996, RNF199, ZCHY, ZNF363
Chromosome Location
Chr.4: 75479037 - 75515057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009922.2 NP_001009922.1 3 - 4 469 137
NM_001278536.1 NP_001265465.1 2 - 3 349 137
NM_001278537.1 NP_001265466.1 2 - 3 349 137
NM_001278538.1 NP_001265467.1 3 - 4 425 137
NM_015436.3 NP_056251.2 3 - 4 469 137
NR_037913.1 3 - 4 469 137
NR_103723.1 3 - 4 469 137
NR_103724.1 3 - 4 469 137
GenBank mRNA AB209072.1 - 3 - 4 332 137
AF247041.1 - 3 - 4 325 137
AF255666.1 - 3 - 4 351 137
AF305424.1 - 3 - 4 359 137
AK091501.1 - 3 - 4 440 137
AK308754.1 - 2 - 3 202 137
AL050144.1 - 2 - 3 228 137
AY888047.1 - 3 - 4 351 137
BC047393.1 - 3 - 4 469 137
BP314571.1 - 2 - 3 216 137
BQ959587.1 - 3 - 4 298 137
DA197338.1 - 3 - 4 425 137
GQ250944.1 - 3 - 4 325 137
GQ250945.1 - 3 - 4 325 137
GQ250946.1 - 3 - 4 325 137
GU937000.1 - 3 - 4 353 137
JF691538.1 - 3 - 4 325 137
JF691539.1 - 3 - 4 325 137

View Full Product Details