The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318246.1 | NP_001305175.1 | 12 - 12 | 3239 | 93 |
NM_194071.3 | NP_919047.2 | 12 - 12 | 3652 | 93 | |
XM_005250541.4 | XP_005250598.1 | 12 - 12 | 4031 | 93 | |
XM_017012540.1 | XP_016868029.1 | 13 - 13 | 5054 | 93 | |
XM_017012541.1 | XP_016868030.1 | 13 - 13 | 3617 | 93 | |
GenBank mRNA | AL080209.1 | - | 1 - 1 | 450 | 93 |
BX648300.1 | - | 1 - 1 | 1484 | 93 | |
BX649143.1 | - | 12 - 12 | 3244 | 93 | |
CR749563.1 | - | 11 - 11 | 3332 | 93 |