Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100521409-100521911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD46
Entrez Gene ID
157567
Gene Name
ankyrin repeat domain 46
Gene Aliases
ANK-S, GENX-115279
UniGene
Hs.530199
Gene Chromosome Location
Chr.8: 100509752-100559786 on Build GRCh38
Transcripts
NM_001270377.1 NM_001270378.1 NM_001270379.1 NM_198401.3 XM_006716509.1 XM_017013106.1

SNP Detail

SNP ID Chromosome Location
rs1010102592 Chr.8: 100521530-100521530 on Build GRCh38
rs931839076 Chr.8: 100521783-100521783 on Build GRCh38
rs890393043 Chr.8: 100521869-100521869 on Build GRCh38
rs186270317 Chr.8: 100521459-100521459 on Build GRCh38
rs190735764 Chr.8: 100521860-100521860 on Build GRCh38
rs1050302730 Chr.8: 100521813-100521813 on Build GRCh38
rs962121556 Chr.8: 100521474-100521474 on Build GRCh38
rs995789732 Chr.8: 100521689-100521689 on Build GRCh38
rs1034030388 Chr.8: 100521634-100521634 on Build GRCh38
rs986963165 Chr.8: 100521829-100521829 on Build GRCh38
rs992800586 Chr.8: 100521617-100521617 on Build GRCh38
rs569452577 Chr.8: 100521718-100521718 on Build GRCh38
rs927552613 Chr.8: 100521599-100521599 on Build GRCh38
rs144900602 Chr.8: 100521508-100521508 on Build GRCh38
rs951617402 Chr.8: 100521819-100521819 on Build GRCh38
rs149024947 Chr.8: 100521534-100521534 on Build GRCh38
rs11998316 Chr.8: 100521498-100521498 on Build GRCh38
rs759891962 Chr.8: 100521618-100521618 on Build GRCh38
rs1043173521 Chr.8: 100521705-100521705 on Build GRCh38
rs796562533 Chr.8: 100521603-100521603 on Build GRCh38
rs1039718144 Chr.8: 100521450-100521450 on Build GRCh38
rs952944308 Chr.8: 100521533-100521533 on Build GRCh38
rs15989 Chr.8: 100521640-100521640 on Build GRCh38
rs753188066 Chr.8: 100521827-100521827 on Build GRCh38
rs539112835 Chr.8: 100521559-100521559 on Build GRCh38
rs556782238 Chr.8: 100521639-100521639 on Build GRCh38
rs772766909 Chr.8: 100521525-100521525 on Build GRCh38
rs959996669 Chr.8: 100521616-100521616 on Build GRCh38
rs1001247827 Chr.8: 100521626-100521626 on Build GRCh38
rs184021756 Chr.8: 100521883-100521883 on Build GRCh38
rs1014654633 Chr.8: 100521467-100521467 on Build GRCh38
rs925767975 Chr.8: 100521720-100521720 on Build GRCh38
rs553645969 Chr.8: 100521496-100521496 on Build GRCh38

View Full Product Details