Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100522143-100522686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD46
Entrez Gene ID
157567
Gene Name
ankyrin repeat domain 46
Gene Aliases
ANK-S, GENX-115279
UniGene
Hs.530199
Gene Chromosome Location
Chr.8: 100509752-100559786 on Build GRCh38
Transcripts
NM_001270377.1 NM_001270378.1 NM_001270379.1 NM_198401.3 XM_006716509.1 XM_017013106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
485876 c.605T>C p.I202T Chr.8: 100522637-100522637 on Build GRCh38
748087 c.637G>A p.V213M Chr.8: 100522605-100522605 on Build GRCh38
4759483 c.662A>T p.E221V Chr.8: 100522580-100522580 on Build GRCh38
4860027 c.636+1G>A p.? Chr.8: 100522605-100522605 on Build GRCh38
485877 c.605T>C p.I202T Chr.8: 100522637-100522637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747335527 Chr.8: 100522513-100522513 on Build GRCh38
rs746100646 Chr.8: 100522310-100522310 on Build GRCh38
rs140363099 Chr.8: 100522336-100522336 on Build GRCh38
rs143185529 Chr.8: 100522181-100522181 on Build GRCh38
rs555690169 Chr.8: 100522383-100522383 on Build GRCh38
rs1044504 Chr.8: 100522524-100522524 on Build GRCh38
rs993062327 Chr.8: 100522174-100522174 on Build GRCh38
rs779157274 Chr.8: 100522288-100522288 on Build GRCh38
rs779832755 Chr.8: 100522492-100522492 on Build GRCh38
rs751985899 Chr.8: 100522642-100522642 on Build GRCh38
rs113970415 Chr.8: 100522536-100522536 on Build GRCh38
rs536765716 Chr.8: 100522351-100522351 on Build GRCh38
rs764351431 Chr.8: 100522554-100522554 on Build GRCh38
rs16898488 Chr.8: 100522505-100522505 on Build GRCh38
rs896015562 Chr.8: 100522451-100522451 on Build GRCh38
rs1025301845 Chr.8: 100522246-100522246 on Build GRCh38
rs967436778 Chr.8: 100522218-100522218 on Build GRCh38
rs1055925913 Chr.8: 100522213-100522213 on Build GRCh38
rs753338330 Chr.8: 100522627-100522627 on Build GRCh38
rs199719979 Chr.8: 100522584-100522584 on Build GRCh38
rs192246523 Chr.8: 100522519-100522519 on Build GRCh38
rs534575713 Chr.8: 100522449-100522449 on Build GRCh38
rs190939994 Chr.8: 100522215-100522215 on Build GRCh38
rs575770928 Chr.8: 100522600-100522600 on Build GRCh38
rs780640944 Chr.8: 100522641-100522641 on Build GRCh38
rs911753732 Chr.8: 100522327-100522327 on Build GRCh38
rs960297460 Chr.8: 100522168-100522168 on Build GRCh38
rs898274679 Chr.8: 100522475-100522475 on Build GRCh38
rs769827601 Chr.8: 100522531-100522531 on Build GRCh38
rs1041304686 Chr.8: 100522532-100522532 on Build GRCh38
rs781612329 Chr.8: 100522651-100522651 on Build GRCh38
rs913863043 Chr.8: 100522397-100522397 on Build GRCh38
rs551008755 Chr.8: 100522348-100522348 on Build GRCh38
rs755447767 Chr.8: 100522647-100522647 on Build GRCh38
rs372015313 Chr.8: 100522550-100522550 on Build GRCh38
rs773309514 Chr.8: 100522534-100522534 on Build GRCh38
rs182854174 Chr.8: 100522546-100522546 on Build GRCh38
rs747566340 Chr.8: 100522562-100522562 on Build GRCh38
rs763554095 Chr.8: 100522606-100522606 on Build GRCh38
rs188125907 Chr.8: 100522256-100522256 on Build GRCh38
rs532422409 Chr.8: 100522252-100522252 on Build GRCh38
rs887400148 Chr.8: 100522289-100522289 on Build GRCh38
rs72679750 Chr.8: 100522421-100522421 on Build GRCh38
rs34603431 Chr.8: 100522595-100522596 on Build GRCh38
rs767173630 Chr.8: 100522561-100522561 on Build GRCh38
rs1033489478 Chr.8: 100522527-100522527 on Build GRCh38
rs1019748792 Chr.8: 100522335-100522335 on Build GRCh38
rs569396967 Chr.8: 100522349-100522349 on Build GRCh38
rs113249311 Chr.8: 100522245-100522245 on Build GRCh38
rs759003595 Chr.8: 100522636-100522636 on Build GRCh38
rs183134328 Chr.8: 100522224-100522224 on Build GRCh38
rs1014449453 Chr.8: 100522455-100522455 on Build GRCh38
rs945952347 Chr.8: 100522405-100522405 on Build GRCh38
rs148262169 Chr.8: 100522311-100522311 on Build GRCh38
rs937090033 Chr.8: 100522207-100522207 on Build GRCh38
rs747652596 Chr.8: 100522596-100522596 on Build GRCh38
rs958363709 Chr.8: 100522569-100522569 on Build GRCh38
rs774165489 Chr.8: 100522547-100522547 on Build GRCh38
rs928473150 Chr.8: 100522422-100522422 on Build GRCh38
rs548740436 Chr.8: 100522365-100522365 on Build GRCh38

View Full Product Details