Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100522440-100522948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD46
Entrez Gene ID
157567
Gene Name
ankyrin repeat domain 46
Gene Aliases
ANK-S, GENX-115279
UniGene
Hs.530199
Gene Chromosome Location
Chr.8: 100509752-100559786 on Build GRCh38
Transcripts
NM_001270377.1 NM_001270378.1 NM_001270379.1 NM_198401.3 XM_006716509.1 XM_017013106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
748087 c.637G>A p.V213M Chr.8: 100522605-100522605 on Build GRCh38
4759483 c.662A>T p.E221V Chr.8: 100522580-100522580 on Build GRCh38
4860027 c.636+1G>A p.? Chr.8: 100522605-100522605 on Build GRCh38
3269157 c.526C>T p.L176F Chr.8: 100522716-100522716 on Build GRCh38
485877 c.605T>C p.I202T Chr.8: 100522637-100522637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757424888 Chr.8: 100522898-100522909 on Build GRCh38
rs747335527 Chr.8: 100522513-100522513 on Build GRCh38
rs750961822 Chr.8: 100522803-100522803 on Build GRCh38
rs770864256 Chr.8: 100522712-100522712 on Build GRCh38
rs931064824 Chr.8: 100522859-100522859 on Build GRCh38
rs372353120 Chr.8: 100522732-100522732 on Build GRCh38
rs187920463 Chr.8: 100522681-100522681 on Build GRCh38
rs1044504 Chr.8: 100522524-100522524 on Build GRCh38
rs943735527 Chr.8: 100522891-100522908 on Build GRCh38
rs774739166 Chr.8: 100522722-100522724 on Build GRCh38
rs7839370 Chr.8: 100522906-100522906 on Build GRCh38
rs549829438 Chr.8: 100522852-100522852 on Build GRCh38
rs35233405 Chr.8: 100522901-100522908 on Build GRCh38
rs779832755 Chr.8: 100522492-100522492 on Build GRCh38
rs199938933 Chr.8: 100522865-100522868 on Build GRCh38
rs751985899 Chr.8: 100522642-100522642 on Build GRCh38
rs16898492 Chr.8: 100522807-100522807 on Build GRCh38
rs113970415 Chr.8: 100522536-100522536 on Build GRCh38
rs1046782325 Chr.8: 100522866-100522866 on Build GRCh38
rs937747474 Chr.8: 100522759-100522759 on Build GRCh38
rs201099342 Chr.8: 100522903-100522908 on Build GRCh38
rs58581117 Chr.8: 100522904-100522909 on Build GRCh38
rs565569412 Chr.8: 100522888-100522888 on Build GRCh38
rs764351431 Chr.8: 100522554-100522554 on Build GRCh38
rs982092867 Chr.8: 100522745-100522745 on Build GRCh38
rs368028062 Chr.8: 100522779-100522779 on Build GRCh38
rs760350477 Chr.8: 100522893-100522910 on Build GRCh38
rs16898488 Chr.8: 100522505-100522505 on Build GRCh38
rs771921223 Chr.8: 100522743-100522743 on Build GRCh38
rs143395176 Chr.8: 100522758-100522758 on Build GRCh38
rs997068698 Chr.8: 100522899-100522906 on Build GRCh38
rs908281514 Chr.8: 100522889-100522908 on Build GRCh38
rs753338330 Chr.8: 100522627-100522627 on Build GRCh38
rs199719979 Chr.8: 100522584-100522584 on Build GRCh38
rs71274981 Chr.8: 100522902-100522909 on Build GRCh38
rs192246523 Chr.8: 100522519-100522519 on Build GRCh38
rs575770928 Chr.8: 100522600-100522600 on Build GRCh38
rs780640944 Chr.8: 100522641-100522641 on Build GRCh38
rs71516869 Chr.8: 100522902-100522902 on Build GRCh38
rs893944274 Chr.8: 100522904-100522905 on Build GRCh38
rs898274679 Chr.8: 100522475-100522475 on Build GRCh38
rs769827601 Chr.8: 100522531-100522531 on Build GRCh38
rs781495158 Chr.8: 100522900-100522907 on Build GRCh38
rs900132359 Chr.8: 100522895-100522908 on Build GRCh38
rs776008084 Chr.8: 100522797-100522797 on Build GRCh38
rs1041304686 Chr.8: 100522532-100522532 on Build GRCh38
rs760295512 Chr.8: 100522789-100522789 on Build GRCh38
rs770363998 Chr.8: 100522903-100522910 on Build GRCh38
rs781612329 Chr.8: 100522651-100522651 on Build GRCh38
rs755447767 Chr.8: 100522647-100522647 on Build GRCh38
rs903456363 Chr.8: 100522884-100522884 on Build GRCh38
rs372015313 Chr.8: 100522550-100522550 on Build GRCh38
rs749790475 Chr.8: 100522901-100522910 on Build GRCh38
rs747171097 Chr.8: 100522894-100522909 on Build GRCh38
rs376494651 Chr.8: 100522865-100522866 on Build GRCh38
rs748356876 Chr.8: 100522672-100522672 on Build GRCh38
rs773309514 Chr.8: 100522534-100522534 on Build GRCh38
rs764908096 Chr.8: 100522899-100522908 on Build GRCh38
rs112984366 Chr.8: 100522907-100522907 on Build GRCh38
rs182854174 Chr.8: 100522546-100522546 on Build GRCh38
rs201248246 Chr.8: 100522904-100522904 on Build GRCh38
rs747566340 Chr.8: 100522562-100522562 on Build GRCh38
rs763554095 Chr.8: 100522606-100522606 on Build GRCh38
rs7839371 Chr.8: 100522908-100522908 on Build GRCh38
rs763761865 Chr.8: 100522796-100522796 on Build GRCh38
rs149022557 Chr.8: 100522686-100522686 on Build GRCh38
rs762285453 Chr.8: 100522746-100522747 on Build GRCh38
rs577749731 Chr.8: 100522703-100522703 on Build GRCh38
rs34603431 Chr.8: 100522595-100522596 on Build GRCh38
rs111598785 Chr.8: 100522910-100522910 on Build GRCh38
rs767173630 Chr.8: 100522561-100522561 on Build GRCh38
rs1033489478 Chr.8: 100522527-100522527 on Build GRCh38
rs745791971 Chr.8: 100522739-100522739 on Build GRCh38
rs530477627 Chr.8: 100522912-100522912 on Build GRCh38
rs758376550 Chr.8: 100522897-100522910 on Build GRCh38
rs192644546 Chr.8: 100522756-100522756 on Build GRCh38
rs59062296 Chr.8: 100522916-100522917 on Build GRCh38
rs1019042072 Chr.8: 100522917-100522917 on Build GRCh38
rs140128819 Chr.8: 100522802-100522803 on Build GRCh38
rs866707384 Chr.8: 100522916-100522916 on Build GRCh38
rs1014375853 Chr.8: 100522862-100522862 on Build GRCh38
rs759003595 Chr.8: 100522636-100522636 on Build GRCh38
rs796258474 Chr.8: 100522906-100522908 on Build GRCh38
rs1000880337 Chr.8: 100522896-100522896 on Build GRCh38
rs1040679065 Chr.8: 100522893-100522908 on Build GRCh38
rs747652596 Chr.8: 100522596-100522596 on Build GRCh38
rs958363709 Chr.8: 100522569-100522569 on Build GRCh38
rs928568088 Chr.8: 100522752-100522752 on Build GRCh38
rs761348378 Chr.8: 100522798-100522798 on Build GRCh38
rs774165489 Chr.8: 100522547-100522547 on Build GRCh38
rs369691090 Chr.8: 100522737-100522737 on Build GRCh38
rs769205440 Chr.8: 100522696-100522696 on Build GRCh38
rs150252002 Chr.8: 100522905-100522906 on Build GRCh38
rs1020928732 Chr.8: 100522864-100522864 on Build GRCh38
rs66638989 Chr.8: 100522905-100522908 on Build GRCh38

View Full Product Details