The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099436.3 | NP_001092906.3 | 4 - 5 | 571 | 74 |
NM_001284364.2 | NP_001271293.2 | 4 - 5 | 571 | 74 | |
NM_001284365.2 | NP_001271294.1 | 4 - 5 | 494 | 74 | |
NR_104307.2 | 4 - 5 | 571 | 74 | ||
NR_104308.2 | 4 - 5 | 571 | 74 | ||
NR_104309.2 | 4 - 5 | 571 | 74 | ||
XM_005254289.2 | XP_005254346.1 | 5 - 6 | 586 | 74 | |
XM_017022068.1 | XP_016877557.1 | 5 - 6 | 620 | 74 | |
XM_017022069.1 | XP_016877558.1 | 5 - 6 | 624 | 74 | |
XM_017022070.1 | XP_016877559.1 | 5 - 6 | 637 | 74 | |
XM_017022071.1 | XP_016877560.1 | 5 - 6 | 624 | 74 | |
XM_017022072.1 | XP_016877561.1 | 4 - 5 | 800 | 74 | |
GenBank mRNA | AK075047.1 | - | 4 - 5 | 480 | 74 |
AK293181.1 | - | 4 - 5 | 424 | 74 | |
AK293650.1 | - | 3 - 4 | 589 | 74 | |
AK294241.1 | - | 4 - 5 | 505 | 74 | |
AK294245.1 | - | 4 - 5 | 501 | 74 | |
AK294251.1 | - | 4 - 5 | 505 | 74 | |
AK298879.1 | - | 4 - 5 | 428 | 74 | |
AK299380.1 | - | 4 - 5 | 498 | 74 | |
AK316102.1 | - | 4 - 5 | 505 | 74 | |
AL117482.1 | - | 4 - 5 | 706 | 74 | |
BC036117.1 | - | 4 - 5 | 542 | 74 | |
BC048289.1 | - | 4 - 5 | 499 | 74 | |
BC056423.1 | - | 4 - 5 | 489 | 74 | |
BC157884.1 | - | 4 - 5 | 553 | 74 | |
BI829133.1 | - | 3 - 4 | 655 | 74 | |
BQ927030.1 | - | 4 - 5 | 459 | 74 |