Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 116938375-116938879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARD
Entrez Gene ID
441376
Gene Name
alanine and arginine rich domain containing protein
Gene Aliases
C8orf85
UniGene
Hs.437551
Gene Chromosome Location
Chr.8: 116937960-116944000 on Build GRCh38
Transcripts
NM_001025357.2 XM_011517043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3395083 c.318G>A p.A106A Chr.8: 116938561-116938561 on Build GRCh38
5976609 c.242_243insGGAGGCGGC p.E81_A82insEAA Chr.8: 116938485-116938486 on Build GRCh38
6051356 c.235G>A p.V79M Chr.8: 116938478-116938478 on Build GRCh38
3698805 c.162G>A p.P54P Chr.8: 116938405-116938405 on Build GRCh38
6047895 c.287G>A p.G96D Chr.8: 116938530-116938530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546125739 Chr.8: 116938760-116938760 on Build GRCh38
rs534177737 Chr.8: 116938497-116938497 on Build GRCh38
rs946959450 Chr.8: 116938538-116938538 on Build GRCh38
rs528285928 Chr.8: 116938848-116938848 on Build GRCh38
rs990969254 Chr.8: 116938805-116938805 on Build GRCh38
rs753093088 Chr.8: 116938460-116938460 on Build GRCh38
rs763439469 Chr.8: 116938468-116938468 on Build GRCh38
rs997067254 Chr.8: 116938785-116938785 on Build GRCh38
rs76753109 Chr.8: 116938453-116938453 on Build GRCh38
rs770410263 Chr.8: 116938493-116938494 on Build GRCh38
rs369425409 Chr.8: 116938500-116938500 on Build GRCh38
rs955525033 Chr.8: 116938592-116938592 on Build GRCh38
rs1044708654 Chr.8: 116938670-116938670 on Build GRCh38
rs755639875 Chr.8: 116938475-116938475 on Build GRCh38
rs745380205 Chr.8: 116938403-116938403 on Build GRCh38
rs961444616 Chr.8: 116938639-116938639 on Build GRCh38
rs948631320 Chr.8: 116938667-116938667 on Build GRCh38
rs951938154 Chr.8: 116938712-116938712 on Build GRCh38
rs768533520 Chr.8: 116938409-116938409 on Build GRCh38
rs140528384 Chr.8: 116938700-116938701 on Build GRCh38
rs938095727 Chr.8: 116938786-116938786 on Build GRCh38
rs900323825 Chr.8: 116938671-116938671 on Build GRCh38
rs946928280 Chr.8: 116938840-116938840 on Build GRCh38
rs372308193 Chr.8: 116938835-116938835 on Build GRCh38
rs939683578 Chr.8: 116938492-116938492 on Build GRCh38
rs772030681 Chr.8: 116938425-116938425 on Build GRCh38
rs776078373 Chr.8: 116938506-116938507 on Build GRCh38
rs777722026 Chr.8: 116938485-116938486 on Build GRCh38
rs367780973 Chr.8: 116938397-116938397 on Build GRCh38
rs760268484 Chr.8: 116938553-116938553 on Build GRCh38
rs1035086073 Chr.8: 116938523-116938523 on Build GRCh38
rs369681281 Chr.8: 116938577-116938577 on Build GRCh38
rs16889284 Chr.8: 116938853-116938853 on Build GRCh38
rs781594426 Chr.8: 116938823-116938824 on Build GRCh38
rs928110895 Chr.8: 116938764-116938764 on Build GRCh38
rs371757231 Chr.8: 116938402-116938402 on Build GRCh38
rs537796659 Chr.8: 116938849-116938850 on Build GRCh38
rs776785256 Chr.8: 116938439-116938439 on Build GRCh38
rs539748650 Chr.8: 116938473-116938473 on Build GRCh38
rs765546287 Chr.8: 116938528-116938528 on Build GRCh38
rs919621012 Chr.8: 116938665-116938665 on Build GRCh38
rs747967818 Chr.8: 116938496-116938496 on Build GRCh38
rs548010933 Chr.8: 116938675-116938675 on Build GRCh38
rs374262247 Chr.8: 116938488-116938488 on Build GRCh38
rs968324793 Chr.8: 116938549-116938549 on Build GRCh38
rs111477672 Chr.8: 116938519-116938519 on Build GRCh38
rs995986299 Chr.8: 116938674-116938674 on Build GRCh38
rs894022089 Chr.8: 116938537-116938537 on Build GRCh38
rs982597747 Chr.8: 116938759-116938759 on Build GRCh38
rs761734653 Chr.8: 116938412-116938412 on Build GRCh38
rs973763986 Chr.8: 116938652-116938652 on Build GRCh38
rs764535166 Chr.8: 116938541-116938541 on Build GRCh38
rs566716899 Chr.8: 116938491-116938491 on Build GRCh38
rs763355279 Chr.8: 116938531-116938531 on Build GRCh38
rs773228226 Chr.8: 116938428-116938428 on Build GRCh38
rs183910056 Chr.8: 116938856-116938856 on Build GRCh38
rs190884662 Chr.8: 116938574-116938574 on Build GRCh38
rs757468249 Chr.8: 116938486-116938488 on Build GRCh38
rs902197205 Chr.8: 116938575-116938575 on Build GRCh38
rs113483821 Chr.8: 116938735-116938735 on Build GRCh38
rs1015810609 Chr.8: 116938829-116938829 on Build GRCh38
rs758416382 Chr.8: 116938481-116938482 on Build GRCh38
rs1015531061 Chr.8: 116938755-116938755 on Build GRCh38
rs747974793 Chr.8: 116938418-116938418 on Build GRCh38
rs908280584 Chr.8: 116938624-116938624 on Build GRCh38
rs187578345 Chr.8: 116938499-116938499 on Build GRCh38
rs983352654 Chr.8: 116938720-116938720 on Build GRCh38
rs751670765 Chr.8: 116938758-116938758 on Build GRCh38
rs752762151 Chr.8: 116938449-116938449 on Build GRCh38
rs1019167634 Chr.8: 116938457-116938457 on Build GRCh38
rs567742710 Chr.8: 116938844-116938844 on Build GRCh38
rs955638520 Chr.8: 116938807-116938807 on Build GRCh38
rs1022739914 Chr.8: 116938548-116938548 on Build GRCh38
rs760711027 Chr.8: 116938429-116938429 on Build GRCh38
rs779888171 Chr.8: 116938480-116938480 on Build GRCh38
rs774353945 Chr.8: 116938414-116938414 on Build GRCh38
rs777740920 Chr.8: 116938498-116938498 on Build GRCh38
rs752076216 Chr.8: 116938551-116938551 on Build GRCh38
rs938228006 Chr.8: 116938526-116938526 on Build GRCh38
rs1018723764 Chr.8: 116938601-116938601 on Build GRCh38
rs542421964 Chr.8: 116938404-116938404 on Build GRCh38
rs766300561 Chr.8: 116938739-116938739 on Build GRCh38
rs375944860 Chr.8: 116938431-116938431 on Build GRCh38
rs892531374 Chr.8: 116938741-116938741 on Build GRCh38
rs575561085 Chr.8: 116938566-116938566 on Build GRCh38
rs986522272 Chr.8: 116938596-116938596 on Build GRCh38
rs759923748 Chr.8: 116938444-116938444 on Build GRCh38
rs16889283 Chr.8: 116938529-116938529 on Build GRCh38
rs970147449 Chr.8: 116938854-116938854 on Build GRCh38
rs1028458915 Chr.8: 116938552-116938552 on Build GRCh38
rs575118752 Chr.8: 116938486-116938491 on Build GRCh38
rs776841268 Chr.8: 116938510-116938510 on Build GRCh38
rs1044230448 Chr.8: 116938518-116938518 on Build GRCh38
rs764630998 Chr.8: 116938471-116938471 on Build GRCh38

View Full Product Details