Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C9orf3
- Entrez Gene ID
-
84909
- Gene Name
- chromosome 9 open reading frame 3
- Gene Aliases
-
AOPEP, AP-O, APO, C90RF3, ONPEP
- Chromosome Location
-
Chr.9: 94726604 - 95148264 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL137535.1 |
- |
1 - 2 |
1347 |
75 |
View Full Product Details