Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KANK1
Entrez Gene ID
23189
Gene Name
KN motif and ankyrin repeat domains 1
Gene Aliases
ANKRD15, CPSQ2, KANK
Chromosome Location
Chr.9: 470294 - 746106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256876.1 NP_001243805.1 7 - 8 3346 100
NM_001256877.1 NP_001243806.1 4 - 5 3455 100
NM_015158.3 NP_055973.2 3 - 4 2840 100
NM_153186.4 NP_694856.1 2 - 3 3102 100
XM_005251411.2 XP_005251468.1 3 - 4 2840 100
XM_005251414.3 XP_005251471.1 2 - 3 3303 100
XM_005251415.2 XP_005251472.1 2 - 3 3032 100
XM_005251416.2 XP_005251473.1 2 - 3 2808 100
XM_005251417.3 XP_005251474.1 2 - 3 3299 100
XM_005251418.2 XP_005251475.1 2 - 3 3310 100
XM_005251419.2 XP_005251476.1 2 - 3 3001 100
XM_006716743.2 XP_006716806.1 3 - 4 2816 100
XM_011517819.1 XP_011516121.1 4 - 5 2931 100
XM_011517820.1 XP_011516122.1 3 - 4 2948 100
XM_011517821.1 XP_011516123.1 3 - 4 2875 100
XM_017014511.1 XP_016870000.1 4 - 5 3053 100
XM_017014512.1 XP_016870001.1 3 - 4 4364 100
XM_017014513.1 XP_016870002.1 5 - 6 3566 100
XM_017014514.1 XP_016870003.1 3 - 4 3252 100
XM_017014515.1 XP_016870004.1 4 - 5 2988 100
XM_017014516.1 XP_016870005.1 3 - 4 3252 100
XM_017014517.1 XP_016870006.1 3 - 4 3252 100
XM_017014518.1 XP_016870007.1 3 - 4 2840 100
XM_017014519.1 XP_016870008.1 3 - 4 3252 100
XM_017014520.1 XP_016870009.1 3 - 4 2840 100
XM_017014521.1 XP_016870010.1 3 - 4 3252 100
XM_017014522.1 XP_016870011.1 4 - 5 3053 100
XM_017014523.1 XP_016870012.1 3 - 4 2840 100
XM_017014524.1 XP_016870013.1 3 - 4 3252 100
XM_017014525.1 XP_016870014.1 3 - 4 3252 100
XM_017014526.1 XP_016870015.1 3 - 4 3285 100
XM_017014527.1 XP_016870016.1 2 - 3 3032 100
XM_017014528.1 XP_016870017.1 3 - 4 2886 100
XM_017014529.1 XP_016870018.1 2 - 3 3299 100
XM_017014530.1 XP_016870019.1 2 - 3 2808 100
XM_017014531.1 XP_016870020.1 2 - 3 3032 100
XM_017014532.1 XP_016870021.1 2 - 3 3306 100
XM_017014533.1 XP_016870022.1 2 - 3 2749 100
XM_017014534.1 XP_016870023.1 2 - 3 3032 100
XM_017014535.1 XP_016870024.1 2 - 3 3298 100
XM_017014536.1 XP_016870025.1 2 - 3 3001 100
XM_017014537.1 XP_016870026.1 2 - 3 2821 100
XM_017014538.1 XP_016870027.1 2 - 3 3306 100
XM_017014539.1 XP_016870028.1 2 - 3 2821 100
XM_017014540.1 XP_016870029.1 2 - 3 3297 100
XM_017014541.1 XP_016870030.1 2 - 3 3301 100
XM_017014542.1 XP_016870031.1 2 - 3 2808 100
GenBank mRNA AK025428.1 - 1 - 2 985 100
AK026421.1 - 1 - 2 2455 100
AK290028.1 - 3 - 4 2806 100
AK292989.1 - 3 - 4 2819 100
AL832454.2 - 3 - 4 2832 100
AL833161.1 - 3 - 4 2820 100
BC020040.1 - 1 - 2 994 100
BC037495.2 - 7 - 8 3349 100
BC038116.2 - 2 - 3 3012 100
D79994.2 - 3 - 4 3405 100

View Full Product Details