Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32405244-32405758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO1
Entrez Gene ID
48
Gene Name
aconitase 1
Gene Aliases
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1
UniGene
Hs.567229
Gene Chromosome Location
Chr.9: 32384603-32450836 on Build GRCh38
Transcripts
NM_001278352.1 NM_002197.2 XM_005251476.1 XM_011517888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4658840 c.56A>C p.K19T Chr.9: 32405562-32405562 on Build GRCh38
3926677 c.15C>T p.F5F Chr.9: 32405521-32405521 on Build GRCh38
5681017 c.82G>T p.E28* Chr.9: 32405588-32405588 on Build GRCh38
4373523 c.72G>A p.L24L Chr.9: 32405578-32405578 on Build GRCh38
6592233 c.17C>T p.A6V Chr.9: 32405523-32405523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41313772 Chr.9: 32405591-32405591 on Build GRCh38
rs987553037 Chr.9: 32405424-32405424 on Build GRCh38
rs373990127 Chr.9: 32405540-32405540 on Build GRCh38
rs774068800 Chr.9: 32405655-32405655 on Build GRCh38
rs769669115 Chr.9: 32405648-32405648 on Build GRCh38
rs748552310 Chr.9: 32405511-32405511 on Build GRCh38
rs1015122227 Chr.9: 32405292-32405292 on Build GRCh38
rs762835739 Chr.9: 32405651-32405651 on Build GRCh38
rs761603433 Chr.9: 32405579-32405579 on Build GRCh38
rs763818525 Chr.9: 32405438-32405438 on Build GRCh38
rs138982398 Chr.9: 32405653-32405653 on Build GRCh38
rs770966790 Chr.9: 32405525-32405525 on Build GRCh38
rs1046384710 Chr.9: 32405647-32405647 on Build GRCh38
rs754371350 Chr.9: 32405488-32405488 on Build GRCh38
rs552946486 Chr.9: 32405611-32405611 on Build GRCh38
rs41298178 Chr.9: 32405641-32405641 on Build GRCh38
rs750981611 Chr.9: 32405450-32405450 on Build GRCh38
rs539136981 Chr.9: 32405437-32405437 on Build GRCh38
rs755480336 Chr.9: 32405594-32405594 on Build GRCh38
rs200697667 Chr.9: 32405650-32405650 on Build GRCh38
rs765711653 Chr.9: 32405273-32405273 on Build GRCh38
rs560315574 Chr.9: 32405569-32405569 on Build GRCh38
rs906802075 Chr.9: 32405275-32405275 on Build GRCh38
rs146405504 Chr.9: 32405380-32405380 on Build GRCh38
rs747211129 Chr.9: 32405627-32405627 on Build GRCh38
rs149388820 Chr.9: 32405710-32405710 on Build GRCh38
rs539573290 Chr.9: 32405361-32405361 on Build GRCh38
rs894985770 Chr.9: 32405672-32405672 on Build GRCh38
rs376599072 Chr.9: 32405563-32405563 on Build GRCh38
rs138295884 Chr.9: 32405528-32405528 on Build GRCh38
rs767118948 Chr.9: 32405580-32405580 on Build GRCh38
rs779491463 Chr.9: 32405598-32405598 on Build GRCh38
rs144056370 Chr.9: 32405341-32405341 on Build GRCh38
rs764889274 Chr.9: 32405336-32405336 on Build GRCh38
rs376224592 Chr.9: 32405497-32405497 on Build GRCh38
rs762827228 Chr.9: 32405564-32405564 on Build GRCh38
rs779419619 Chr.9: 32405491-32405491 on Build GRCh38
rs977439368 Chr.9: 32405517-32405517 on Build GRCh38
rs899402791 Chr.9: 32405702-32405702 on Build GRCh38
rs571682590 Chr.9: 32405426-32405426 on Build GRCh38
rs552325935 Chr.9: 32405338-32405339 on Build GRCh38
rs771208100 Chr.9: 32405634-32405634 on Build GRCh38
rs752959345 Chr.9: 32405607-32405607 on Build GRCh38
rs939665183 Chr.9: 32405628-32405628 on Build GRCh38
rs953128926 Chr.9: 32405394-32405394 on Build GRCh38
rs963543398 Chr.9: 32405321-32405321 on Build GRCh38
rs759995241 Chr.9: 32405269-32405269 on Build GRCh38
rs756857226 Chr.9: 32405472-32405472 on Build GRCh38
rs781182402 Chr.9: 32405639-32405639 on Build GRCh38
rs1033959112 Chr.9: 32405369-32405369 on Build GRCh38
rs868355818 Chr.9: 32405681-32405681 on Build GRCh38
rs370326617 Chr.9: 32405618-32405618 on Build GRCh38
rs1005131152 Chr.9: 32405276-32405276 on Build GRCh38
rs759464257 Chr.9: 32405544-32405544 on Build GRCh38
rs772130307 Chr.9: 32405521-32405521 on Build GRCh38
rs755604412 Chr.9: 32405490-32405490 on Build GRCh38
rs906313462 Chr.9: 32405339-32405339 on Build GRCh38
rs201900147 Chr.9: 32405522-32405522 on Build GRCh38
rs1031755059 Chr.9: 32405523-32405523 on Build GRCh38
rs1036785184 Chr.9: 32405334-32405334 on Build GRCh38
rs916664302 Chr.9: 32405602-32405602 on Build GRCh38
rs751190138 Chr.9: 32405572-32405572 on Build GRCh38
rs374959721 Chr.9: 32405310-32405310 on Build GRCh38

View Full Product Details