Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32418245-32418740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO1
Entrez Gene ID
48
Gene Name
aconitase 1
Gene Aliases
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1
UniGene
Hs.567229
Gene Chromosome Location
Chr.9: 32384603-32450836 on Build GRCh38
Transcripts
NM_001278352.1 NM_002197.2 XM_005251476.1 XM_011517888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5583241 c.488C>T p.A163V Chr.9: 32418341-32418341 on Build GRCh38
3703538 c.530T>C p.I177T Chr.9: 32418383-32418383 on Build GRCh38
4919207 c.499A>G p.M167V Chr.9: 32418352-32418352 on Build GRCh38
3906787 c.501G>A p.M167I Chr.9: 32418354-32418354 on Build GRCh38
4373536 c.565G>T p.V189L Chr.9: 32418418-32418418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879141690 Chr.9: 32418277-32418277 on Build GRCh38
rs766152770 Chr.9: 32418379-32418379 on Build GRCh38
rs768708549 Chr.9: 32418446-32418446 on Build GRCh38
rs373332582 Chr.9: 32418311-32418311 on Build GRCh38
rs778052946 Chr.9: 32418293-32418293 on Build GRCh38
rs904683578 Chr.9: 32418272-32418272 on Build GRCh38
rs571342824 Chr.9: 32418657-32418657 on Build GRCh38
rs934245773 Chr.9: 32418585-32418585 on Build GRCh38
rs1036093594 Chr.9: 32418457-32418457 on Build GRCh38
rs749679991 Chr.9: 32418432-32418432 on Build GRCh38
rs888997704 Chr.9: 32418478-32418478 on Build GRCh38
rs368519878 Chr.9: 32418531-32418531 on Build GRCh38
rs544613435 Chr.9: 32418362-32418362 on Build GRCh38
rs749513351 Chr.9: 32418351-32418351 on Build GRCh38
rs111674920 Chr.9: 32418530-32418530 on Build GRCh38
rs139223145 Chr.9: 32418456-32418456 on Build GRCh38
rs371287650 Chr.9: 32418418-32418418 on Build GRCh38
rs774234584 Chr.9: 32418356-32418356 on Build GRCh38
rs1053715202 Chr.9: 32418564-32418564 on Build GRCh38
rs770259136 Chr.9: 32418298-32418298 on Build GRCh38
rs780495634 Chr.9: 32418427-32418427 on Build GRCh38
rs764759386 Chr.9: 32418527-32418527 on Build GRCh38
rs756219806 Chr.9: 32418526-32418527 on Build GRCh38
rs145069089 Chr.9: 32418689-32418689 on Build GRCh38
rs143260336 Chr.9: 32418388-32418388 on Build GRCh38
rs901386343 Chr.9: 32418583-32418583 on Build GRCh38
rs2274721 Chr.9: 32418279-32418279 on Build GRCh38
rs1028410753 Chr.9: 32418330-32418330 on Build GRCh38
rs201517506 Chr.9: 32418499-32418499 on Build GRCh38
rs377699256 Chr.9: 32418411-32418411 on Build GRCh38
rs371235086 Chr.9: 32418471-32418471 on Build GRCh38
rs756628451 Chr.9: 32418549-32418549 on Build GRCh38
rs759044295 Chr.9: 32418391-32418391 on Build GRCh38
rs61753543 Chr.9: 32418475-32418475 on Build GRCh38
rs776354014 Chr.9: 32418497-32418497 on Build GRCh38
rs760657982 Chr.9: 32418371-32418371 on Build GRCh38
rs772171891 Chr.9: 32418474-32418474 on Build GRCh38
rs746415598 Chr.9: 32418295-32418295 on Build GRCh38
rs771866685 Chr.9: 32418365-32418365 on Build GRCh38
rs750952732 Chr.9: 32418420-32418420 on Build GRCh38
rs530451988 Chr.9: 32418421-32418421 on Build GRCh38
rs41305321 Chr.9: 32418355-32418355 on Build GRCh38
rs760606540 Chr.9: 32418485-32418485 on Build GRCh38
rs1031023465 Chr.9: 32418611-32418611 on Build GRCh38
rs770860316 Chr.9: 32418491-32418491 on Build GRCh38
rs200294382 Chr.9: 32418539-32418539 on Build GRCh38
rs750556772 Chr.9: 32418474-32418474 on Build GRCh38
rs940448475 Chr.9: 32418409-32418409 on Build GRCh38
rs763718428 Chr.9: 32418598-32418598 on Build GRCh38
rs774021644 Chr.9: 32418674-32418674 on Build GRCh38
rs1018767705 Chr.9: 32418580-32418580 on Build GRCh38
rs545140455 Chr.9: 32418292-32418292 on Build GRCh38
rs185730378 Chr.9: 32418294-32418294 on Build GRCh38
rs771023115 Chr.9: 32418328-32418328 on Build GRCh38
rs763691738 Chr.9: 32418538-32418538 on Build GRCh38
rs561386458 Chr.9: 32418417-32418417 on Build GRCh38
rs773106532 Chr.9: 32418366-32418366 on Build GRCh38
rs752101251 Chr.9: 32418397-32418397 on Build GRCh38
rs573577461 Chr.9: 32418636-32418636 on Build GRCh38
rs551143278 Chr.9: 32418595-32418595 on Build GRCh38
rs776053905 Chr.9: 32418350-32418350 on Build GRCh38
rs146705506 Chr.9: 32418453-32418453 on Build GRCh38
rs200265624 Chr.9: 32418314-32418314 on Build GRCh38

View Full Product Details