Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32425714-32426258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO1
Entrez Gene ID
48
Gene Name
aconitase 1
Gene Aliases
ACONS, HEL60, IREB1, IREBP, IREBP1, IRP1
UniGene
Hs.567229
Gene Chromosome Location
Chr.9: 32384603-32450836 on Build GRCh38
Transcripts
NM_001278352.1 NM_002197.2 XM_005251476.1 XM_011517888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3763877 c.1263A>G p.E421E Chr.9: 32425912-32425912 on Build GRCh38
1108118 c.1328C>T p.P443L Chr.9: 32425977-32425977 on Build GRCh38
1108121 c.1342G>A p.G448R Chr.9: 32425991-32425991 on Build GRCh38
3763876 c.1263A>G p.E421E Chr.9: 32425912-32425912 on Build GRCh38
4752292 c.1343G>T p.G448V Chr.9: 32425992-32425992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752903469 Chr.9: 32426044-32426044 on Build GRCh38
rs759790204 Chr.9: 32426038-32426038 on Build GRCh38
rs567203838 Chr.9: 32425877-32425877 on Build GRCh38
rs913218770 Chr.9: 32426199-32426199 on Build GRCh38
rs1003779874 Chr.9: 32426062-32426062 on Build GRCh38
rs529985830 Chr.9: 32425889-32425889 on Build GRCh38
rs1027323252 Chr.9: 32426103-32426103 on Build GRCh38
rs991893011 Chr.9: 32426121-32426121 on Build GRCh38
rs545042295 Chr.9: 32425810-32425810 on Build GRCh38
rs770781100 Chr.9: 32425970-32425970 on Build GRCh38
rs917737191 Chr.9: 32425822-32425822 on Build GRCh38
rs748250321 Chr.9: 32425858-32425858 on Build GRCh38
rs554173248 Chr.9: 32426166-32426166 on Build GRCh38
rs757787003 Chr.9: 32425801-32425801 on Build GRCh38
rs1018507649 Chr.9: 32426081-32426081 on Build GRCh38
rs540880410 Chr.9: 32426117-32426117 on Build GRCh38
rs763742544 Chr.9: 32425908-32425908 on Build GRCh38
rs112535632 Chr.9: 32425978-32425978 on Build GRCh38
rs201745454 Chr.9: 32425927-32425927 on Build GRCh38
rs746948322 Chr.9: 32425960-32425960 on Build GRCh38
rs756475720 Chr.9: 32425817-32425817 on Build GRCh38
rs976358555 Chr.9: 32425820-32425820 on Build GRCh38
rs138520929 Chr.9: 32425853-32425853 on Build GRCh38
rs991943673 Chr.9: 32425812-32425812 on Build GRCh38
rs143851438 Chr.9: 32426095-32426095 on Build GRCh38
rs776361289 Chr.9: 32425879-32425879 on Build GRCh38
rs775256231 Chr.9: 32425988-32425988 on Build GRCh38
rs766815541 Chr.9: 32426025-32426025 on Build GRCh38
rs564853182 Chr.9: 32425826-32425826 on Build GRCh38
rs910433762 Chr.9: 32425777-32425777 on Build GRCh38
rs762519258 Chr.9: 32425905-32425905 on Build GRCh38
rs755158738 Chr.9: 32425844-32425844 on Build GRCh38
rs780340742 Chr.9: 32425837-32425837 on Build GRCh38
rs761183852 Chr.9: 32426018-32426018 on Build GRCh38
rs1051100858 Chr.9: 32426234-32426234 on Build GRCh38
rs772543828 Chr.9: 32425821-32425822 on Build GRCh38
rs113600182 Chr.9: 32425955-32425955 on Build GRCh38
rs199591033 Chr.9: 32425899-32425899 on Build GRCh38
rs749227184 Chr.9: 32425866-32425866 on Build GRCh38
rs376056668 Chr.9: 32426052-32426052 on Build GRCh38
rs769668610 Chr.9: 32425984-32425984 on Build GRCh38
rs1057066581 Chr.9: 32426068-32426068 on Build GRCh38
rs776431987 Chr.9: 32425977-32425977 on Build GRCh38
rs951119110 Chr.9: 32425793-32425793 on Build GRCh38
rs946080855 Chr.9: 32426219-32426219 on Build GRCh38
rs182949449 Chr.9: 32425933-32425933 on Build GRCh38
rs560785479 Chr.9: 32425749-32425749 on Build GRCh38
rs778214990 Chr.9: 32425833-32425833 on Build GRCh38
rs746998837 Chr.9: 32425875-32425875 on Build GRCh38
rs531082463 Chr.9: 32425791-32425791 on Build GRCh38
rs773102797 Chr.9: 32425872-32425872 on Build GRCh38
rs530391630 Chr.9: 32425841-32425841 on Build GRCh38
rs969222670 Chr.9: 32426162-32426162 on Build GRCh38
rs936577549 Chr.9: 32426193-32426193 on Build GRCh38
rs34319839 Chr.9: 32425900-32425900 on Build GRCh38
rs139154946 Chr.9: 32425943-32425943 on Build GRCh38
rs915263030 Chr.9: 32425917-32425917 on Build GRCh38
rs946667312 Chr.9: 32425922-32425922 on Build GRCh38
rs754192532 Chr.9: 32426032-32426032 on Build GRCh38
rs976012617 Chr.9: 32426232-32426232 on Build GRCh38
rs951721247 Chr.9: 32426107-32426107 on Build GRCh38
rs140192396 Chr.9: 32426078-32426078 on Build GRCh38
rs115097215 Chr.9: 32426198-32426198 on Build GRCh38
rs765639080 Chr.9: 32426042-32426042 on Build GRCh38
rs777891541 Chr.9: 32425949-32425949 on Build GRCh38
rs766698567 Chr.9: 32425915-32425915 on Build GRCh38
rs563068608 Chr.9: 32425774-32425774 on Build GRCh38
rs750948884 Chr.9: 32425910-32425910 on Build GRCh38
rs1051149996 Chr.9: 32426047-32426047 on Build GRCh38
rs1007213058 Chr.9: 32426114-32426114 on Build GRCh38
rs1017266693 Chr.9: 32426189-32426189 on Build GRCh38
rs755178170 Chr.9: 32425932-32425932 on Build GRCh38
rs965980216 Chr.9: 32425786-32425786 on Build GRCh38
rs375525370 Chr.9: 32425762-32425762 on Build GRCh38
rs768276333 Chr.9: 32425998-32425998 on Build GRCh38
rs1024740729 Chr.9: 32426123-32426123 on Build GRCh38
rs774002493 Chr.9: 32426015-32426015 on Build GRCh38
rs963291727 Chr.9: 32426083-32426083 on Build GRCh38
rs1007485659 Chr.9: 32426059-32426059 on Build GRCh38
rs762749604 Chr.9: 32425997-32425997 on Build GRCh38
rs751003420 Chr.9: 32425811-32425811 on Build GRCh38
rs550196915 Chr.9: 32425867-32425867 on Build GRCh38
rs3780473 Chr.9: 32425912-32425912 on Build GRCh38

View Full Product Details