Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33402378-33402889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP7
Entrez Gene ID
364
Gene Name
aquaporin 7
Gene Aliases
AQP7L, AQPap, GLYCQTL
UniGene
Hs.455323
Gene Chromosome Location
Chr.9: 33383135-33402682 on Build GRCh38
Transcripts
NM_001170.2 NM_001318156.1 NM_001318157.1 NM_001318158.1 NR_134513.1 NR_134514.1 NR_134515.1 XM_005251453.3 XM_011517866.2 XM_017014700.1 XM_017014701.1 XM_017014702.1 XM_017014705.1 XM_017014706.1

SNP Detail

SNP ID Chromosome Location
rs66774754 Chr.9: 33402551-33402551 on Build GRCh38
rs755036690 Chr.9: 33402529-33402529 on Build GRCh38
rs573072525 Chr.9: 33402446-33402446 on Build GRCh38
rs753915934 Chr.9: 33402833-33402833 on Build GRCh38
rs12003531 Chr.9: 33402650-33402650 on Build GRCh38
rs980115483 Chr.9: 33402575-33402575 on Build GRCh38
rs536926889 Chr.9: 33402845-33402845 on Build GRCh38
rs1051491783 Chr.9: 33402645-33402645 on Build GRCh38
rs375508425 Chr.9: 33402541-33402541 on Build GRCh38
rs67030231 Chr.9: 33402442-33402442 on Build GRCh38
rs890820954 Chr.9: 33402445-33402445 on Build GRCh38
rs971645951 Chr.9: 33402768-33402768 on Build GRCh38
rs980530469 Chr.9: 33402790-33402790 on Build GRCh38
rs868069246 Chr.9: 33402548-33402548 on Build GRCh38
rs936064953 Chr.9: 33402587-33402587 on Build GRCh38
rs556952557 Chr.9: 33402856-33402856 on Build GRCh38
rs1005031600 Chr.9: 33402691-33402691 on Build GRCh38
rs1025389587 Chr.9: 33402670-33402670 on Build GRCh38
rs551593232 Chr.9: 33402676-33402676 on Build GRCh38
rs1018469729 Chr.9: 33402450-33402450 on Build GRCh38
rs148220051 Chr.9: 33402501-33402502 on Build GRCh38
rs957460606 Chr.9: 33402857-33402857 on Build GRCh38
rs777882139 Chr.9: 33402570-33402570 on Build GRCh38
rs12003509 Chr.9: 33402653-33402653 on Build GRCh38
rs530244834 Chr.9: 33402552-33402552 on Build GRCh38
rs62544589 Chr.9: 33402555-33402555 on Build GRCh38
rs12005633 Chr.9: 33402738-33402738 on Build GRCh38
rs760878067 Chr.9: 33402425-33402425 on Build GRCh38
rs12003516 Chr.9: 33402593-33402593 on Build GRCh38
rs879110181 Chr.9: 33402828-33402828 on Build GRCh38
rs1006556206 Chr.9: 33402449-33402449 on Build GRCh38
rs945875285 Chr.9: 33402644-33402644 on Build GRCh38
rs76444615 Chr.9: 33402620-33402620 on Build GRCh38
rs898249629 Chr.9: 33402667-33402667 on Build GRCh38
rs12003564 Chr.9: 33402854-33402854 on Build GRCh38
rs565197184 Chr.9: 33402532-33402532 on Build GRCh38
rs1039347413 Chr.9: 33402665-33402665 on Build GRCh38
rs898218496 Chr.9: 33402475-33402475 on Build GRCh38
rs367801575 Chr.9: 33402863-33402863 on Build GRCh38
rs995021761 Chr.9: 33402480-33402480 on Build GRCh38
rs984141453 Chr.9: 33402540-33402540 on Build GRCh38
rs927385075 Chr.9: 33402582-33402582 on Build GRCh38
rs913317101 Chr.9: 33402866-33402866 on Build GRCh38
rs547089351 Chr.9: 33402654-33402654 on Build GRCh38
rs184021017 Chr.9: 33402488-33402488 on Build GRCh38
rs144044323 Chr.9: 33402436-33402436 on Build GRCh38
rs62544590 Chr.9: 33402780-33402780 on Build GRCh38
rs377531213 Chr.9: 33402669-33402669 on Build GRCh38
rs766293019 Chr.9: 33402434-33402434 on Build GRCh38
rs12003520 Chr.9: 33402714-33402714 on Build GRCh38
rs62544588 Chr.9: 33402531-33402531 on Build GRCh38
rs369833349 Chr.9: 33402543-33402543 on Build GRCh38
rs528257956 Chr.9: 33402675-33402675 on Build GRCh38
rs62544591 Chr.9: 33402797-33402797 on Build GRCh38

View Full Product Details